Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32288947-32289172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDL3
Entrez Gene ID
162494
Gene Name
rhomboid like 3
Gene Aliases
RHBDL4, VRHO
UniGene
Hs.655027
Gene Chromosome Location
Chr.17: 32266167-32324661 on Build GRCh38
Transcripts
NM_138328.2 XM_006721733.3 XM_006721734.3 XM_011524443.2 XM_017024272.1 XM_017024273.1 XM_017024274.1 XM_017024275.1 XM_017024276.1 XM_017024277.1 XM_017024278.1 XM_017024279.1 XM_017024280.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6766134 c.480delC p.P162fs*5 Chr.17: 32288977-32288977 on Build GRCh38
3180339 c.509C>T p.T170M Chr.17: 32289006-32289006 on Build GRCh38
3370669 c.459C>T p.Y153Y Chr.17: 32288956-32288956 on Build GRCh38
5386036 c.493T>C p.F165L Chr.17: 32288990-32288990 on Build GRCh38
4065400 c.510G>A p.T170T Chr.17: 32289007-32289007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763183414 Chr.17: 32289034-32289035 on Build GRCh38
rs932564279 Chr.17: 32289073-32289073 on Build GRCh38
rs763183513 Chr.17: 32288981-32288981 on Build GRCh38
rs769268879 Chr.17: 32288979-32288979 on Build GRCh38
rs773658861 Chr.17: 32289013-32289013 on Build GRCh38
rs768973472 Chr.17: 32288993-32288993 on Build GRCh38
rs561086057 Chr.17: 32289123-32289123 on Build GRCh38
rs374589254 Chr.17: 32289024-32289024 on Build GRCh38
rs1055100593 Chr.17: 32289088-32289088 on Build GRCh38
rs751469103 Chr.17: 32289065-32289065 on Build GRCh38
rs774745615 Chr.17: 32288997-32288997 on Build GRCh38
rs141556431 Chr.17: 32289134-32289134 on Build GRCh38
rs148353778 Chr.17: 32289107-32289107 on Build GRCh38
rs768077438 Chr.17: 32289007-32289007 on Build GRCh38
rs926806844 Chr.17: 32289122-32289122 on Build GRCh38
rs750402314 Chr.17: 32289014-32289014 on Build GRCh38
rs547014704 Chr.17: 32289098-32289098 on Build GRCh38
rs563403864 Chr.17: 32288982-32288982 on Build GRCh38
rs565282405 Chr.17: 32289006-32289006 on Build GRCh38
rs987909571 Chr.17: 32289069-32289069 on Build GRCh38
rs945635823 Chr.17: 32289034-32289034 on Build GRCh38
rs780847434 Chr.17: 32288972-32288972 on Build GRCh38
rs199643919 Chr.17: 32289063-32289063 on Build GRCh38
rs980114897 Chr.17: 32289040-32289040 on Build GRCh38
rs914126969 Chr.17: 32289027-32289027 on Build GRCh38
rs1004261477 Chr.17: 32288973-32288973 on Build GRCh38
rs922494233 Chr.17: 32289066-32289066 on Build GRCh38
rs371750736 Chr.17: 32289022-32289022 on Build GRCh38
rs755300624 Chr.17: 32289030-32289030 on Build GRCh38
rs530345368 Chr.17: 32289079-32289079 on Build GRCh38
rs561120912 Chr.17: 32289070-32289070 on Build GRCh38

View Full Product Details