Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44756412-44756639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMIZ2
Entrez Gene ID
83637
Gene Name
zinc finger MIZ-type containing 2
Gene Aliases
NET27, TRAFIP20, ZIMP7, hZIMP7
UniGene
Hs.600681
Gene Chromosome Location
Chr.7: 44748547-44769881 on Build GRCh38
Transcripts
NM_001300959.1 NM_031449.3 NM_174929.2 XM_005249866.2 XM_005249867.4 XM_005249868.4 XM_005249869.2 XM_005249870.2 XM_005249871.2 XM_005249872.2 XM_005249873.2 XM_017012674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3263039 c.114G>A p.S38S Chr.7: 44756488-44756488 on Build GRCh38
73383 c.157C>G p.Q53E Chr.7: 44756531-44756531 on Build GRCh38
421434 c.63C>T p.F21F Chr.7: 44756437-44756437 on Build GRCh38
4187204 c.152C>T p.A51V Chr.7: 44756526-44756526 on Build GRCh38
5407562 c.113C>T p.S38L Chr.7: 44756487-44756487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758983021 Chr.7: 44756544-44756544 on Build GRCh38
rs1010503123 Chr.7: 44756613-44756613 on Build GRCh38
rs905642358 Chr.7: 44756535-44756535 on Build GRCh38
rs773134661 Chr.7: 44756577-44756585 on Build GRCh38
rs755700642 Chr.7: 44756562-44756562 on Build GRCh38
rs780567663 Chr.7: 44756556-44756556 on Build GRCh38
rs201771017 Chr.7: 44756452-44756452 on Build GRCh38
rs201139620 Chr.7: 44756440-44756440 on Build GRCh38
rs374086780 Chr.7: 44756439-44756439 on Build GRCh38
rs762226810 Chr.7: 44756525-44756525 on Build GRCh38
rs200119008 Chr.7: 44756447-44756447 on Build GRCh38
rs759501345 Chr.7: 44756565-44756565 on Build GRCh38
rs746553504 Chr.7: 44756579-44756582 on Build GRCh38
rs530185242 Chr.7: 44756533-44756533 on Build GRCh38
rs369543914 Chr.7: 44756574-44756574 on Build GRCh38
rs771915502 Chr.7: 44756574-44756575 on Build GRCh38
rs763445027 Chr.7: 44756526-44756526 on Build GRCh38
rs375678046 Chr.7: 44756524-44756524 on Build GRCh38
rs918274345 Chr.7: 44756519-44756519 on Build GRCh38
rs201213111 Chr.7: 44756437-44756437 on Build GRCh38
rs759480693 Chr.7: 44756479-44756479 on Build GRCh38
rs559541332 Chr.7: 44756527-44756527 on Build GRCh38
rs893994977 Chr.7: 44756438-44756438 on Build GRCh38
rs769722298 Chr.7: 44756566-44756566 on Build GRCh38
rs201936829 Chr.7: 44756567-44756567 on Build GRCh38
rs772072381 Chr.7: 44756488-44756488 on Build GRCh38
rs770653755 Chr.7: 44756571-44756571 on Build GRCh38
rs764422172 Chr.7: 44756523-44756523 on Build GRCh38
rs563630212 Chr.7: 44756616-44756616 on Build GRCh38
rs752310033 Chr.7: 44756559-44756559 on Build GRCh38
rs749158165 Chr.7: 44756450-44756450 on Build GRCh38
rs552291020 Chr.7: 44756595-44756595 on Build GRCh38
rs760795097 Chr.7: 44756501-44756501 on Build GRCh38
rs775705219 Chr.7: 44756494-44756494 on Build GRCh38
rs943996738 Chr.7: 44756456-44756456 on Build GRCh38
rs747959510 Chr.7: 44756573-44756587 on Build GRCh38
rs772054359 Chr.7: 44756575-44756575 on Build GRCh38
rs199684927 Chr.7: 44756468-44756468 on Build GRCh38

View Full Product Details