Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119032828-119033055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WARS2
Entrez Gene ID
10352
Gene Name
tryptophanyl tRNA synthetase 2, mitochondrial
Gene Aliases
TrpRS
UniGene
Hs.523506
Gene Chromosome Location
Chr.1: 119031216-119140701 on Build GRCh38
Transcripts
NM_015836.3 NM_201263.2 XM_005270350.3 XM_011540494.2 XM_011540495.2 XM_017000038.1 XM_017000039.1 XM_017000040.1 XM_017000041.1 XM_017000042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
462731 c.947C>G p.P316R Chr.1: 119033047-119033047 on Build GRCh38
3472174 c.1020A>G p.A340A Chr.1: 119032974-119032974 on Build GRCh38
3472173 c.1057G>A p.V353M Chr.1: 119032937-119032937 on Build GRCh38
4021231 c.1031A>C p.E344A Chr.1: 119032963-119032963 on Build GRCh38
364904 c.1018G>C p.A340P Chr.1: 119032976-119032976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199858756 Chr.1: 119032877-119032877 on Build GRCh38
rs751126372 Chr.1: 119033021-119033021 on Build GRCh38
rs780674665 Chr.1: 119033024-119033024 on Build GRCh38
rs949484894 Chr.1: 119032968-119032968 on Build GRCh38
rs780398419 Chr.1: 119032866-119032866 on Build GRCh38
rs151047248 Chr.1: 119033031-119033031 on Build GRCh38
rs774787983 Chr.1: 119032873-119032873 on Build GRCh38
rs770370630 Chr.1: 119032862-119032862 on Build GRCh38
rs369838673 Chr.1: 119032878-119032878 on Build GRCh38
rs563341344 Chr.1: 119032940-119032940 on Build GRCh38
rs759396511 Chr.1: 119032923-119032923 on Build GRCh38
rs931308856 Chr.1: 119032937-119032937 on Build GRCh38
rs777998766 Chr.1: 119032937-119032939 on Build GRCh38
rs766389848 Chr.1: 119032904-119032904 on Build GRCh38
rs373768460 Chr.1: 119032900-119032900 on Build GRCh38
rs1046598440 Chr.1: 119033006-119033006 on Build GRCh38
rs774567512 Chr.1: 119032864-119032864 on Build GRCh38
rs17023101 Chr.1: 119032915-119032915 on Build GRCh38
rs773372731 Chr.1: 119032899-119032899 on Build GRCh38
rs916656828 Chr.1: 119032967-119032967 on Build GRCh38
rs758094787 Chr.1: 119032977-119032977 on Build GRCh38
rs752435347 Chr.1: 119032942-119032942 on Build GRCh38
rs201923721 Chr.1: 119032871-119032871 on Build GRCh38
rs28641023 Chr.1: 119032924-119032924 on Build GRCh38
rs754288860 Chr.1: 119032965-119032965 on Build GRCh38
rs762139228 Chr.1: 119032874-119032874 on Build GRCh38
rs777256506 Chr.1: 119033017-119033017 on Build GRCh38
rs756706541 Chr.1: 119033022-119033022 on Build GRCh38

View Full Product Details