Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18431328-18431591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP4
Entrez Gene ID
170463
Gene Name
single stranded DNA binding protein 4
Gene Aliases
-
UniGene
Hs.515259
Gene Chromosome Location
Chr.19: 18418719-18434562 on Build GRCh38
Transcripts
NM_001009998.3 NM_032627.4 XM_005259790.3 XM_006722665.3 XM_006722666.2 XM_006722668.2 XM_017026431.1 XM_017026432.1 XM_017026433.1 XM_017026434.1 XM_017026435.1 XM_017026436.1 XM_017026437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3530806 c.396C>T p.P132P Chr.19: 18431379-18431379 on Build GRCh38
1391678 c.372delC p.G127fs*13 Chr.19: 18431355-18431355 on Build GRCh38
4491771 c.385C>T p.Q129* Chr.19: 18431368-18431368 on Build GRCh38
6465630 c.420G>A p.M140I Chr.19: 18431403-18431403 on Build GRCh38
1288203 c.408C>T p.N136N Chr.19: 18431391-18431391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183851202 Chr.19: 18431373-18431373 on Build GRCh38
rs778945918 Chr.19: 18431431-18431431 on Build GRCh38
rs941172794 Chr.19: 18431418-18431418 on Build GRCh38
rs990230176 Chr.19: 18431543-18431543 on Build GRCh38
rs754231661 Chr.19: 18431386-18431386 on Build GRCh38
rs753163077 Chr.19: 18431393-18431393 on Build GRCh38
rs959158315 Chr.19: 18431469-18431469 on Build GRCh38
rs924619885 Chr.19: 18431521-18431521 on Build GRCh38
rs539511836 Chr.19: 18431406-18431406 on Build GRCh38
rs761436464 Chr.19: 18431370-18431370 on Build GRCh38
rs966670070 Chr.19: 18431486-18431486 on Build GRCh38
rs545959018 Chr.19: 18431538-18431538 on Build GRCh38
rs766718404 Chr.19: 18431380-18431380 on Build GRCh38
rs755343600 Chr.19: 18431444-18431445 on Build GRCh38
rs1040580131 Chr.19: 18431564-18431564 on Build GRCh38
rs544171112 Chr.19: 18431555-18431555 on Build GRCh38
rs575881522 Chr.19: 18431540-18431540 on Build GRCh38
rs774101701 Chr.19: 18431362-18431362 on Build GRCh38
rs377300360 Chr.19: 18431440-18431440 on Build GRCh38
rs566676951 Chr.19: 18431454-18431454 on Build GRCh38
rs960354693 Chr.19: 18431549-18431549 on Build GRCh38
rs868575669 Chr.19: 18431537-18431537 on Build GRCh38
rs768646934 Chr.19: 18431358-18431358 on Build GRCh38
rs887464334 Chr.19: 18431438-18431438 on Build GRCh38
rs771763234 Chr.19: 18431441-18431441 on Build GRCh38
rs553057980 Chr.19: 18431412-18431412 on Build GRCh38
rs1009184690 Chr.19: 18431473-18431473 on Build GRCh38
rs910644163 Chr.19: 18431551-18431551 on Build GRCh38
rs867503763 Chr.19: 18431359-18431359 on Build GRCh38
rs530924478 Chr.19: 18431390-18431390 on Build GRCh38
rs900695998 Chr.19: 18431480-18431480 on Build GRCh38
rs751838128 Chr.19: 18431408-18431408 on Build GRCh38
rs535721917 Chr.19: 18431467-18431467 on Build GRCh38
rs199685072 Chr.19: 18431398-18431398 on Build GRCh38
rs771673414 Chr.19: 18431371-18431371 on Build GRCh38
rs766959351 Chr.19: 18431354-18431355 on Build GRCh38
rs920678372 Chr.19: 18431565-18431565 on Build GRCh38
rs760883618 Chr.19: 18431355-18431355 on Build GRCh38
rs867451595 Chr.19: 18431387-18431387 on Build GRCh38
rs978435547 Chr.19: 18431519-18431519 on Build GRCh38
rs779158437 Chr.19: 18431443-18431444 on Build GRCh38
rs773045873 Chr.19: 18431372-18431372 on Build GRCh38
rs772843090 Chr.19: 18431446-18431446 on Build GRCh38
rs369515073 Chr.19: 18431470-18431470 on Build GRCh38
rs1054572149 Chr.19: 18431346-18431346 on Build GRCh38
rs1025282813 Chr.19: 18431475-18431475 on Build GRCh38
rs754426399 Chr.19: 18431438-18431438 on Build GRCh38
rs556013452 Chr.19: 18431505-18431505 on Build GRCh38
rs374446728 Chr.19: 18431410-18431410 on Build GRCh38
rs770630427 Chr.19: 18431460-18431460 on Build GRCh38
rs746206379 Chr.19: 18431522-18431522 on Build GRCh38
rs113198578 Chr.19: 18431391-18431391 on Build GRCh38
rs751157126 Chr.19: 18431454-18431455 on Build GRCh38
rs200775325 Chr.19: 18431413-18431413 on Build GRCh38
rs943485071 Chr.19: 18431561-18431561 on Build GRCh38

View Full Product Details