Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119383017-119383290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP31
Entrez Gene ID
57514
Gene Name
Rho GTPase activating protein 31
Gene Aliases
AOS1, CDGAP
UniGene
Hs.668218
Gene Chromosome Location
Chr.3: 119294289-119419476 on Build GRCh38
Transcripts
NM_020754.3 XM_006713714.3 XM_017006955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036690 c.594C>T p.V198V Chr.3: 119383138-119383138 on Build GRCh38
5048879 c.542C>G p.S181C Chr.3: 119383086-119383086 on Build GRCh38
1036691 c.613_614insT p.E206fs*1 Chr.3: 119383157-119383158 on Build GRCh38
5570902 c.611T>C p.V204A Chr.3: 119383155-119383155 on Build GRCh38
5756148 c.610G>A p.V204M Chr.3: 119383154-119383154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768208725 Chr.3: 119383151-119383151 on Build GRCh38
rs369993733 Chr.3: 119383071-119383071 on Build GRCh38
rs770123367 Chr.3: 119383052-119383052 on Build GRCh38
rs753559800 Chr.3: 119383175-119383175 on Build GRCh38
rs763977018 Chr.3: 119383253-119383253 on Build GRCh38
rs769060777 Chr.3: 119383216-119383216 on Build GRCh38
rs761116153 Chr.3: 119383169-119383169 on Build GRCh38
rs1051603058 Chr.3: 119383055-119383055 on Build GRCh38
rs368795933 Chr.3: 119383158-119383158 on Build GRCh38
rs145326425 Chr.3: 119383198-119383198 on Build GRCh38
rs774626312 Chr.3: 119383142-119383142 on Build GRCh38
rs771296646 Chr.3: 119383241-119383241 on Build GRCh38
rs773793308 Chr.3: 119383235-119383235 on Build GRCh38
rs759183962 Chr.3: 119383246-119383246 on Build GRCh38
rs898795745 Chr.3: 119383056-119383056 on Build GRCh38
rs766740253 Chr.3: 119383141-119383141 on Build GRCh38
rs764713087 Chr.3: 119383177-119383177 on Build GRCh38
rs777537877 Chr.3: 119383121-119383121 on Build GRCh38
rs775801076 Chr.3: 119383135-119383135 on Build GRCh38
rs748245130 Chr.3: 119383223-119383223 on Build GRCh38
rs1010784829 Chr.3: 119383183-119383183 on Build GRCh38
rs777932793 Chr.3: 119383106-119383106 on Build GRCh38
rs539898181 Chr.3: 119383080-119383080 on Build GRCh38
rs759160533 Chr.3: 119383176-119383176 on Build GRCh38
rs202118332 Chr.3: 119383220-119383220 on Build GRCh38
rs758387698 Chr.3: 119383119-119383119 on Build GRCh38
rs199650901 Chr.3: 119383103-119383103 on Build GRCh38
rs370881248 Chr.3: 119383199-119383199 on Build GRCh38
rs772651005 Chr.3: 119383233-119383233 on Build GRCh38
rs765900101 Chr.3: 119383171-119383171 on Build GRCh38
rs760909492 Chr.3: 119383239-119383239 on Build GRCh38
rs758262903 Chr.3: 119383185-119383185 on Build GRCh38
rs72966429 Chr.3: 119383245-119383245 on Build GRCh38
rs764873014 Chr.3: 119383249-119383249 on Build GRCh38
rs768829713 Chr.3: 119383140-119383140 on Build GRCh38
rs749283093 Chr.3: 119383139-119383139 on Build GRCh38
rs373737244 Chr.3: 119383250-119383250 on Build GRCh38
rs748055794 Chr.3: 119383117-119383117 on Build GRCh38
rs147532572 Chr.3: 119383038-119383039 on Build GRCh38
rs775025186 Chr.3: 119383077-119383077 on Build GRCh38
rs762400554 Chr.3: 119383150-119383150 on Build GRCh38
rs753231280 Chr.3: 119383110-119383110 on Build GRCh38
rs751793 Chr.3: 119383206-119383206 on Build GRCh38
rs746065771 Chr.3: 119383037-119383037 on Build GRCh38
rs375920880 Chr.3: 119383059-119383059 on Build GRCh38
rs746822864 Chr.3: 119383124-119383124 on Build GRCh38
rs769848017 Chr.3: 119383125-119383125 on Build GRCh38
rs764455668 Chr.3: 119383073-119383073 on Build GRCh38
rs777699046 Chr.3: 119383189-119383189 on Build GRCh38
rs375159706 Chr.3: 119383041-119383042 on Build GRCh38
rs376381685 Chr.3: 119383111-119383111 on Build GRCh38
rs762469484 Chr.3: 119383251-119383251 on Build GRCh38
rs754008141 Chr.3: 119383184-119383184 on Build GRCh38
rs16829782 Chr.3: 119383078-119383078 on Build GRCh38
rs751260180 Chr.3: 119383197-119383197 on Build GRCh38
rs943629668 Chr.3: 119383041-119383041 on Build GRCh38

View Full Product Details