Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48731231-48731494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDFY4
Entrez Gene ID
57705
Gene Name
WDFY family member 4
Gene Aliases
C10orf64
UniGene
Hs.287379
Gene Chromosome Location
Chr.10: 48684815-48982956 on Build GRCh38
Transcripts
NM_020945.1 XM_005270004.3 XM_011539986.2 XM_011539987.2 XM_011539988.2 XM_011539990.2 XM_011539991.2 XM_011539992.2 XM_017016463.1 XM_017016465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910360 c.1397T>C p.I466T Chr.10: 48731377-48731377 on Build GRCh38
918503 c.1253G>A p.R418Q Chr.10: 48731233-48731233 on Build GRCh38
7317512 c.1479C>T p.I493I Chr.10: 48731459-48731459 on Build GRCh38
5078022 c.1320G>A p.S440S Chr.10: 48731273-48731273 on Build GRCh38
918505 c.1489G>C p.D497H Chr.10: 48731469-48731469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74358986 Chr.10: 48731428-48731428 on Build GRCh38
rs1055574215 Chr.10: 48731264-48731264 on Build GRCh38
rs372273417 Chr.10: 48731417-48731417 on Build GRCh38
rs995170652 Chr.10: 48731372-48731372 on Build GRCh38
rs865793313 Chr.10: 48731340-48731340 on Build GRCh38
rs778872914 Chr.10: 48731383-48731383 on Build GRCh38
rs752971235 Chr.10: 48731363-48731363 on Build GRCh38
rs368232022 Chr.10: 48731302-48731302 on Build GRCh38
rs572346860 Chr.10: 48731312-48731312 on Build GRCh38
rs766845809 Chr.10: 48731307-48731307 on Build GRCh38
rs897450600 Chr.10: 48731309-48731309 on Build GRCh38
rs765774619 Chr.10: 48731460-48731460 on Build GRCh38
rs186660788 Chr.10: 48731273-48731273 on Build GRCh38
rs78148589 Chr.10: 48731427-48731427 on Build GRCh38
rs557285079 Chr.10: 48731393-48731393 on Build GRCh38
rs574912591 Chr.10: 48731444-48731444 on Build GRCh38
rs746512306 Chr.10: 48731426-48731426 on Build GRCh38
rs759602707 Chr.10: 48731331-48731331 on Build GRCh38
rs1013855364 Chr.10: 48731420-48731420 on Build GRCh38
rs865977690 Chr.10: 48731289-48731289 on Build GRCh38
rs868418330 Chr.10: 48731369-48731369 on Build GRCh38
rs918541681 Chr.10: 48731285-48731285 on Build GRCh38
rs200340504 Chr.10: 48731303-48731303 on Build GRCh38
rs76028336 Chr.10: 48731430-48731430 on Build GRCh38
rs749693808 Chr.10: 48731464-48731464 on Build GRCh38
rs1007112656 Chr.10: 48731328-48731328 on Build GRCh38
rs35365900 Chr.10: 48731293-48731293 on Build GRCh38
rs368931313 Chr.10: 48731459-48731459 on Build GRCh38
rs767767458 Chr.10: 48731351-48731351 on Build GRCh38
rs771466567 Chr.10: 48731472-48731472 on Build GRCh38
rs773533795 Chr.10: 48731423-48731423 on Build GRCh38
rs1018349076 Chr.10: 48731434-48731434 on Build GRCh38
rs371231718 Chr.10: 48731405-48731405 on Build GRCh38
rs372095585 Chr.10: 48731362-48731362 on Build GRCh38
rs539358549 Chr.10: 48731382-48731382 on Build GRCh38
rs535669602 Chr.10: 48731294-48731294 on Build GRCh38
rs779920041 Chr.10: 48731425-48731425 on Build GRCh38
rs79099016 Chr.10: 48731429-48731429 on Build GRCh38

View Full Product Details