Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11947040-11947279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF69
Entrez Gene ID
7620
Gene Name
zinc finger protein 69
Gene Aliases
Cos5, hZNF3
UniGene
Hs.565280
Gene Chromosome Location
Chr.19: 11887773-11980222 on Build GRCh38
Transcripts
XM_017027231.1 NM_001271848.1 NM_144566.2

Gene Symbol
ZNF700
Entrez Gene ID
90592
Gene Name
zinc finger protein 700
Gene Aliases
-
UniGene
Hs.528486
Gene Chromosome Location
Chr.19: 11925068-11950773 on Build GRCh38
Transcripts
XM_017027231.1 NM_001271848.1 NM_144566.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6897421 c.151G>A p.E51K Chr.19: 11947268-11947268 on Build GRCh38
3959539 c.79G>A p.E27K Chr.19: 11947196-11947196 on Build GRCh38
3728404 c.91G>A p.V31M Chr.19: 11947208-11947208 on Build GRCh38
1390467 c.154G>C p.V52L Chr.19: 11947271-11947271 on Build GRCh38
6341978 c.117A>T p.T39T Chr.19: 11947234-11947234 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530134452 Chr.19: 11947157-11947157 on Build GRCh38
rs1002801609 Chr.19: 11947120-11947120 on Build GRCh38
rs544037747 Chr.19: 11947063-11947063 on Build GRCh38
rs754619442 Chr.19: 11947208-11947208 on Build GRCh38
rs759830729 Chr.19: 11947122-11947122 on Build GRCh38
rs761817983 Chr.19: 11947165-11947165 on Build GRCh38
rs758263083 Chr.19: 11947187-11947187 on Build GRCh38
rs751225821 Chr.19: 11947201-11947201 on Build GRCh38
rs559868416 Chr.19: 11947117-11947117 on Build GRCh38
rs1020991550 Chr.19: 11947190-11947190 on Build GRCh38
rs58336952 Chr.19: 11947101-11947101 on Build GRCh38
rs780731557 Chr.19: 11947209-11947209 on Build GRCh38
rs372080793 Chr.19: 11947093-11947093 on Build GRCh38
rs767940434 Chr.19: 11947134-11947134 on Build GRCh38
rs776730597 Chr.19: 11947236-11947236 on Build GRCh38
rs780051954 Chr.19: 11947193-11947193 on Build GRCh38
rs769194963 Chr.19: 11947222-11947222 on Build GRCh38
rs998183128 Chr.19: 11947213-11947213 on Build GRCh38
rs976330915 Chr.19: 11947180-11947180 on Build GRCh38
rs775680256 Chr.19: 11947144-11947144 on Build GRCh38
rs570568275 Chr.19: 11947172-11947172 on Build GRCh38
rs770910372 Chr.19: 11947235-11947235 on Build GRCh38
rs150476749 Chr.19: 11947221-11947221 on Build GRCh38
rs565478825 Chr.19: 11947079-11947079 on Build GRCh38
rs777674708 Chr.19: 11947086-11947086 on Build GRCh38
rs772407320 Chr.19: 11947228-11947228 on Build GRCh38
rs548287051 Chr.19: 11947225-11947225 on Build GRCh38
rs750389227 Chr.19: 11947181-11947181 on Build GRCh38
rs764046186 Chr.19: 11947163-11947163 on Build GRCh38
rs918888210 Chr.19: 11947081-11947081 on Build GRCh38
rs760975029 Chr.19: 11947234-11947234 on Build GRCh38
rs775868311 Chr.19: 11947230-11947230 on Build GRCh38

View Full Product Details