Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41943630-41943891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM33
Entrez Gene ID
55161
Gene Name
transmembrane protein 33
Gene Aliases
1600019D15Rik, SHINC-3, SHINC3
UniGene
Hs.31082
Gene Chromosome Location
Chr.4: 41935120-41960807 on Build GRCh38
Transcripts
NM_018126.2 XM_005248116.3 XM_005248117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954057 c.380C>T p.T127M Chr.4: 41943798-41943798 on Build GRCh38
6804010 c.360G>A p.L120L Chr.4: 41943778-41943778 on Build GRCh38
1055430 c.396+1G>A p.? Chr.4: 41943815-41943815 on Build GRCh38
6384682 c.339C>A p.F113L Chr.4: 41943757-41943757 on Build GRCh38
5502853 c.396+3A>T p.? Chr.4: 41943817-41943817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916467432 Chr.4: 41943675-41943675 on Build GRCh38
rs772677833 Chr.4: 41943831-41943832 on Build GRCh38
rs765652422 Chr.4: 41943801-41943801 on Build GRCh38
rs753017641 Chr.4: 41943805-41943805 on Build GRCh38
rs751698978 Chr.4: 41943829-41943829 on Build GRCh38
rs758941055 Chr.4: 41943807-41943807 on Build GRCh38
rs754211754 Chr.4: 41943713-41943713 on Build GRCh38
rs746115814 Chr.4: 41943849-41943849 on Build GRCh38
rs551179337 Chr.4: 41943758-41943758 on Build GRCh38
rs767718252 Chr.4: 41943798-41943798 on Build GRCh38
rs760247773 Chr.4: 41943787-41943787 on Build GRCh38
rs751905788 Chr.4: 41943749-41943749 on Build GRCh38
rs749108113 Chr.4: 41943764-41943764 on Build GRCh38
rs370877822 Chr.4: 41943784-41943784 on Build GRCh38
rs772096586 Chr.4: 41943783-41943783 on Build GRCh38
rs575043047 Chr.4: 41943693-41943693 on Build GRCh38
rs771110292 Chr.4: 41943754-41943754 on Build GRCh38
rs375945633 Chr.4: 41943731-41943731 on Build GRCh38
rs1008893984 Chr.4: 41943845-41943845 on Build GRCh38
rs549137721 Chr.4: 41943660-41943660 on Build GRCh38
rs571073263 Chr.4: 41943769-41943769 on Build GRCh38
rs766012828 Chr.4: 41943792-41943792 on Build GRCh38
rs777858612 Chr.4: 41943733-41943733 on Build GRCh38
rs978944682 Chr.4: 41943725-41943725 on Build GRCh38
rs1028919920 Chr.4: 41943806-41943806 on Build GRCh38
rs755364547 Chr.4: 41943714-41943714 on Build GRCh38
rs781070668 Chr.4: 41943841-41943841 on Build GRCh38
rs750594350 Chr.4: 41943827-41943827 on Build GRCh38
rs185525877 Chr.4: 41943717-41943717 on Build GRCh38
rs748120295 Chr.4: 41943773-41943773 on Build GRCh38
rs116070412 Chr.4: 41943796-41943796 on Build GRCh38
rs374683858 Chr.4: 41943720-41943720 on Build GRCh38
rs746217017 Chr.4: 41943737-41943737 on Build GRCh38
rs764564535 Chr.4: 41943811-41943811 on Build GRCh38
rs890005352 Chr.4: 41943815-41943815 on Build GRCh38
rs757358054 Chr.4: 41943834-41943834 on Build GRCh38
rs893581074 Chr.4: 41943793-41943793 on Build GRCh38
rs762118434 Chr.4: 41943765-41943765 on Build GRCh38
rs747057995 Chr.4: 41943748-41943748 on Build GRCh38
rs374153513 Chr.4: 41943856-41943856 on Build GRCh38
rs774231768 Chr.4: 41943771-41943771 on Build GRCh38

View Full Product Details