Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25686296-25686533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF337
Entrez Gene ID
26152
Gene Name
zinc finger protein 337
Gene Aliases
-
UniGene
Hs.61881
Gene Chromosome Location
Chr.20: 25673195-25696904 on Build GRCh38
Transcripts
NM_001290261.1 NM_015655.3 XM_005260702.3 XM_006723558.3 XM_011529219.2 XM_017027802.1 XM_017027803.1 XM_017027804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
337079 c.18C>A p.A6A Chr.20: 25686400-25686400 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200266867 Chr.20: 25686401-25686401 on Build GRCh38
rs759135011 Chr.20: 25686431-25686431 on Build GRCh38
rs754888590 Chr.20: 25686461-25686461 on Build GRCh38
rs184069633 Chr.20: 25686370-25686370 on Build GRCh38
rs941822033 Chr.20: 25686490-25686490 on Build GRCh38
rs878940368 Chr.20: 25686500-25686500 on Build GRCh38
rs910376647 Chr.20: 25686475-25686475 on Build GRCh38
rs199528333 Chr.20: 25686494-25686494 on Build GRCh38
rs749861679 Chr.20: 25686419-25686419 on Build GRCh38
rs903135847 Chr.20: 25686374-25686374 on Build GRCh38
rs189102189 Chr.20: 25686438-25686438 on Build GRCh38
rs1003564746 Chr.20: 25686405-25686405 on Build GRCh38
rs753675137 Chr.20: 25686439-25686439 on Build GRCh38
rs1034504996 Chr.20: 25686421-25686421 on Build GRCh38
rs766306409 Chr.20: 25686365-25686365 on Build GRCh38
rs772937948 Chr.20: 25686347-25686347 on Build GRCh38
rs377383107 Chr.20: 25686403-25686403 on Build GRCh38
rs975216980 Chr.20: 25686449-25686449 on Build GRCh38
rs752216113 Chr.20: 25686381-25686381 on Build GRCh38
rs777429831 Chr.20: 25686386-25686386 on Build GRCh38
rs984166760 Chr.20: 25686478-25686478 on Build GRCh38
rs967104924 Chr.20: 25686446-25686446 on Build GRCh38
rs193013587 Chr.20: 25686465-25686465 on Build GRCh38
rs778927125 Chr.20: 25686432-25686432 on Build GRCh38
rs747400638 Chr.20: 25686348-25686348 on Build GRCh38
rs550054940 Chr.20: 25686391-25686391 on Build GRCh38
rs773354451 Chr.20: 25686444-25686444 on Build GRCh38
rs750824750 Chr.20: 25686387-25686387 on Build GRCh38
rs377355685 Chr.20: 25686447-25686447 on Build GRCh38
rs929919811 Chr.20: 25686328-25686328 on Build GRCh38
rs1030426380 Chr.20: 25686450-25686450 on Build GRCh38
rs570628073 Chr.20: 25686353-25686353 on Build GRCh38
rs894674642 Chr.20: 25686427-25686427 on Build GRCh38
rs753733034 Chr.20: 25686369-25686369 on Build GRCh38
rs373132665 Chr.20: 25686377-25686377 on Build GRCh38
rs145736120 Chr.20: 25686496-25686497 on Build GRCh38
rs758049022 Chr.20: 25686382-25686382 on Build GRCh38

View Full Product Details