Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123416649-123416888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDYHV1
Entrez Gene ID
55093
Gene Name
WDYHV motif containing 1
Gene Aliases
C8orf32
UniGene
Hs.18029
Gene Chromosome Location
Chr.8: 123416725-123467527 on Build GRCh38
Transcripts
NM_001283024.1 NM_001283027.1 NM_018024.2 NR_133926.1 XM_006716597.3 XM_011517147.1 XM_011517151.2 XM_017013601.1 XM_017013602.1 XM_017013603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5979591 c.19G>A p.A7T Chr.8: 123416868-123416868 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888469291 Chr.8: 123416685-123416685 on Build GRCh38
rs750854560 Chr.8: 123416851-123416851 on Build GRCh38
rs1050654020 Chr.8: 123416728-123416741 on Build GRCh38
rs751815494 Chr.8: 123416788-123416788 on Build GRCh38
rs866194175 Chr.8: 123416705-123416705 on Build GRCh38
rs955374363 Chr.8: 123416698-123416698 on Build GRCh38
rs112754744 Chr.8: 123416731-123416731 on Build GRCh38
rs949619878 Chr.8: 123416796-123416796 on Build GRCh38
rs950015749 Chr.8: 123416809-123416809 on Build GRCh38
rs1008676065 Chr.8: 123416720-123416720 on Build GRCh38
rs961792487 Chr.8: 123416730-123416730 on Build GRCh38
rs959506267 Chr.8: 123416735-123416735 on Build GRCh38
rs371588266 Chr.8: 123416868-123416868 on Build GRCh38
rs1030231528 Chr.8: 123416688-123416688 on Build GRCh38
rs945798323 Chr.8: 123416747-123416747 on Build GRCh38
rs761855800 Chr.8: 123416808-123416808 on Build GRCh38
rs35231447 Chr.8: 123416709-123416710 on Build GRCh38
rs900357208 Chr.8: 123416864-123416864 on Build GRCh38
rs556762019 Chr.8: 123416686-123416686 on Build GRCh38
rs991045721 Chr.8: 123416775-123416775 on Build GRCh38
rs996098537 Chr.8: 123416687-123416687 on Build GRCh38
rs35662393 Chr.8: 123416851-123416852 on Build GRCh38
rs879292531 Chr.8: 123416797-123416797 on Build GRCh38
rs989131740 Chr.8: 123416804-123416804 on Build GRCh38
rs1033655344 Chr.8: 123416683-123416683 on Build GRCh38
rs368459108 Chr.8: 123416726-123416726 on Build GRCh38
rs868859933 Chr.8: 123416794-123416794 on Build GRCh38
rs536875179 Chr.8: 123416672-123416672 on Build GRCh38
rs925609071 Chr.8: 123416782-123416782 on Build GRCh38
rs936426317 Chr.8: 123416825-123416825 on Build GRCh38
rs568499775 Chr.8: 123416834-123416834 on Build GRCh38
rs961934991 Chr.8: 123416719-123416719 on Build GRCh38
rs931776644 Chr.8: 123416866-123416866 on Build GRCh38
rs888435923 Chr.8: 123416871-123416871 on Build GRCh38
rs1045560383 Chr.8: 123416826-123416827 on Build GRCh38
rs756647782 Chr.8: 123416855-123416855 on Build GRCh38
rs1015888712 Chr.8: 123416714-123416714 on Build GRCh38
rs918196542 Chr.8: 123416778-123416778 on Build GRCh38
rs187779135 Chr.8: 123416848-123416848 on Build GRCh38
rs923624653 Chr.8: 123416815-123416815 on Build GRCh38
rs920093157 Chr.8: 123416734-123416734 on Build GRCh38
rs559553089 Chr.8: 123416733-123416733 on Build GRCh38
rs913109005 Chr.8: 123416805-123416805 on Build GRCh38

View Full Product Details