Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 228016506-228016775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPHKAP
Entrez Gene ID
80309
Gene Name
SPHK1 interactor, AKAP domain containing
Gene Aliases
SKIP
UniGene
Hs.436306
Gene Chromosome Location
Chr.2: 227979954-228181999 on Build GRCh38
Transcripts
NM_001142644.1 NM_030623.3 XM_005246870.4 XM_006712777.3 XM_006712778.3 XM_011511925.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442146 c.4161G>C p.L1387F Chr.2: 228016693-228016693 on Build GRCh38
3578583 c.4324G>A p.E1442K Chr.2: 228016530-228016530 on Build GRCh38
4493594 c.4181C>T p.T1394I Chr.2: 228016673-228016673 on Build GRCh38
3578585 c.4321G>A p.G1441R Chr.2: 228016533-228016533 on Build GRCh38
4493644 c.4192C>T p.P1398S Chr.2: 228016662-228016662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143578745 Chr.2: 228016658-228016658 on Build GRCh38
rs767125740 Chr.2: 228016563-228016563 on Build GRCh38
rs963208885 Chr.2: 228016691-228016691 on Build GRCh38
rs139756256 Chr.2: 228016551-228016551 on Build GRCh38
rs768713763 Chr.2: 228016723-228016723 on Build GRCh38
rs750035282 Chr.2: 228016576-228016576 on Build GRCh38
rs760207640 Chr.2: 228016676-228016676 on Build GRCh38
rs370817362 Chr.2: 228016531-228016531 on Build GRCh38
rs144013612 Chr.2: 228016585-228016585 on Build GRCh38
rs780266628 Chr.2: 228016714-228016714 on Build GRCh38
rs779020319 Chr.2: 228016732-228016732 on Build GRCh38
rs752670685 Chr.2: 228016705-228016705 on Build GRCh38
rs766082431 Chr.2: 228016679-228016679 on Build GRCh38
rs747714256 Chr.2: 228016722-228016722 on Build GRCh38
rs151068684 Chr.2: 228016633-228016633 on Build GRCh38
rs190144340 Chr.2: 228016602-228016602 on Build GRCh38
rs370387532 Chr.2: 228016735-228016735 on Build GRCh38
rs772667973 Chr.2: 228016742-228016742 on Build GRCh38
rs762176818 Chr.2: 228016553-228016553 on Build GRCh38
rs535121231 Chr.2: 228016712-228016712 on Build GRCh38
rs749391388 Chr.2: 228016623-228016623 on Build GRCh38
rs143055592 Chr.2: 228016749-228016749 on Build GRCh38
rs753344811 Chr.2: 228016680-228016680 on Build GRCh38
rs755848234 Chr.2: 228016582-228016582 on Build GRCh38
rs1004561185 Chr.2: 228016645-228016645 on Build GRCh38
rs752474867 Chr.2: 228016599-228016599 on Build GRCh38
rs757731200 Chr.2: 228016728-228016728 on Build GRCh38
rs200100138 Chr.2: 228016666-228016666 on Build GRCh38
rs774839623 Chr.2: 228016634-228016634 on Build GRCh38
rs773668965 Chr.2: 228016664-228016664 on Build GRCh38
rs867066923 Chr.2: 228016617-228016617 on Build GRCh38
rs917966530 Chr.2: 228016628-228016628 on Build GRCh38
rs748343264 Chr.2: 228016626-228016627 on Build GRCh38
rs764615338 Chr.2: 228016535-228016535 on Build GRCh38
rs945499421 Chr.2: 228016635-228016635 on Build GRCh38
rs937996175 Chr.2: 228016614-228016614 on Build GRCh38
rs993255240 Chr.2: 228016729-228016730 on Build GRCh38
rs144202259 Chr.2: 228016747-228016747 on Build GRCh38
rs746053623 Chr.2: 228016609-228016609 on Build GRCh38
rs867443313 Chr.2: 228016644-228016644 on Build GRCh38
rs913204735 Chr.2: 228016731-228016731 on Build GRCh38
rs758411799 Chr.2: 228016709-228016709 on Build GRCh38
rs373331468 Chr.2: 228016600-228016600 on Build GRCh38
rs764908088 Chr.2: 228016704-228016704 on Build GRCh38
rs926024344 Chr.2: 228016746-228016746 on Build GRCh38
rs751398719 Chr.2: 228016711-228016711 on Build GRCh38
rs952364970 Chr.2: 228016561-228016561 on Build GRCh38
rs770092990 Chr.2: 228016616-228016616 on Build GRCh38
rs985002513 Chr.2: 228016571-228016571 on Build GRCh38
rs1029914968 Chr.2: 228016624-228016624 on Build GRCh38
rs550054274 Chr.2: 228016590-228016590 on Build GRCh38
rs2396519 Chr.2: 228016598-228016598 on Build GRCh38
rs760739252 Chr.2: 228016586-228016586 on Build GRCh38
rs762446477 Chr.2: 228016640-228016640 on Build GRCh38
rs139455126 Chr.2: 228016741-228016741 on Build GRCh38
rs780067739 Chr.2: 228016621-228016621 on Build GRCh38
rs759122353 Chr.2: 228016688-228016688 on Build GRCh38
rs370848705 Chr.2: 228016534-228016534 on Build GRCh38
rs377022345 Chr.2: 228016710-228016710 on Build GRCh38

View Full Product Details