Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101727184-101727443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
UniGene
Hs.444135
Gene Chromosome Location
Chr.3: 101724098-101770562 on Build GRCh38
Transcripts
NM_001303401.1 NM_024548.3 XM_006713743.3 XM_011513127.1 XM_017007178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7261173 c.205C>T p.R69W Chr.3: 101727401-101727401 on Build GRCh38
5476408 c.193G>A p.V65I Chr.3: 101727389-101727389 on Build GRCh38
5509953 c.215G>A p.R72Q Chr.3: 101727411-101727411 on Build GRCh38
6255440 c.240G>T p.T80T Chr.3: 101727436-101727436 on Build GRCh38
5559933 c.209T>A p.L70Q Chr.3: 101727405-101727405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897020666 Chr.3: 101727300-101727300 on Build GRCh38
rs757656908 Chr.3: 101727381-101727381 on Build GRCh38
rs1016370346 Chr.3: 101727374-101727374 on Build GRCh38
rs901766953 Chr.3: 101727307-101727307 on Build GRCh38
rs776962234 Chr.3: 101727378-101727378 on Build GRCh38
rs759268940 Chr.3: 101727350-101727350 on Build GRCh38
rs150064368 Chr.3: 101727260-101727260 on Build GRCh38
rs757981286 Chr.3: 101727369-101727369 on Build GRCh38
rs375541863 Chr.3: 101727384-101727384 on Build GRCh38
rs753533975 Chr.3: 101727343-101727343 on Build GRCh38
rs72584959 Chr.3: 101727365-101727366 on Build GRCh38
rs752129117 Chr.3: 101727380-101727380 on Build GRCh38
rs866202332 Chr.3: 101727255-101727255 on Build GRCh38
rs547567941 Chr.3: 101727410-101727410 on Build GRCh38
rs746188103 Chr.3: 101727402-101727402 on Build GRCh38
rs3070526 Chr.3: 101727364-101727365 on Build GRCh38
rs1044417417 Chr.3: 101727252-101727252 on Build GRCh38
rs552210320 Chr.3: 101727327-101727327 on Build GRCh38
rs780133671 Chr.3: 101727408-101727408 on Build GRCh38
rs368575291 Chr.3: 101727364-101727364 on Build GRCh38
rs997965605 Chr.3: 101727342-101727342 on Build GRCh38
rs756408387 Chr.3: 101727407-101727407 on Build GRCh38
rs997466465 Chr.3: 101727309-101727309 on Build GRCh38
rs563667340 Chr.3: 101727239-101727239 on Build GRCh38
rs941081403 Chr.3: 101727275-101727278 on Build GRCh38
rs571119048 Chr.3: 101727336-101727336 on Build GRCh38
rs139328680 Chr.3: 101727401-101727401 on Build GRCh38
rs1041614424 Chr.3: 101727303-101727303 on Build GRCh38
rs397806871 Chr.3: 101727366-101727367 on Build GRCh38
rs916829021 Chr.3: 101727226-101727226 on Build GRCh38
rs764771645 Chr.3: 101727362-101727362 on Build GRCh38
rs774363921 Chr.3: 101727420-101727420 on Build GRCh38
rs760547308 Chr.3: 101727332-101727332 on Build GRCh38
rs775821951 Chr.3: 101727219-101727219 on Build GRCh38
rs200184476 Chr.3: 101727288-101727291 on Build GRCh38
rs948408499 Chr.3: 101727229-101727229 on Build GRCh38
rs760927923 Chr.3: 101727221-101727221 on Build GRCh38

View Full Product Details