Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72288395-72288648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF6
Entrez Gene ID
60677
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
BRUNOL6
UniGene
Hs.348342
Gene Chromosome Location
Chr.15: 72284727-72320184 on Build GRCh38
Transcripts
NM_001172684.1 NM_001172685.1 NM_052840.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1937320 c.1097C>A p.A366D Chr.15: 72288615-72288615 on Build GRCh38
4522535 c.828G>A p.Q276Q Chr.15: 72288545-72288545 on Build GRCh38
4826043 c.828G>C p.Q276H Chr.15: 72288545-72288545 on Build GRCh38
5667472 c.1229C>A p.A410E Chr.15: 72288397-72288397 on Build GRCh38
4056686 c.1214A>G p.Q405R Chr.15: 72288412-72288412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760982790 Chr.15: 72288522-72288523 on Build GRCh38
rs762962128 Chr.15: 72288428-72288428 on Build GRCh38
rs371909592 Chr.15: 72288489-72288489 on Build GRCh38
rs939019908 Chr.15: 72288485-72288485 on Build GRCh38
rs185544168 Chr.15: 72288458-72288458 on Build GRCh38
rs754254022 Chr.15: 72288576-72288576 on Build GRCh38
rs373381789 Chr.15: 72288543-72288543 on Build GRCh38
rs761929086 Chr.15: 72288446-72288446 on Build GRCh38
rs760953520 Chr.15: 72288556-72288556 on Build GRCh38
rs368776944 Chr.15: 72288527-72288527 on Build GRCh38
rs780863728 Chr.15: 72288606-72288606 on Build GRCh38
rs374383155 Chr.15: 72288499-72288499 on Build GRCh38
rs768614123 Chr.15: 72288522-72288522 on Build GRCh38
rs773360805 Chr.15: 72288551-72288551 on Build GRCh38
rs777101200 Chr.15: 72288475-72288475 on Build GRCh38
rs756814573 Chr.15: 72288600-72288600 on Build GRCh38
rs767694867 Chr.15: 72288447-72288447 on Build GRCh38
rs376352620 Chr.15: 72288560-72288560 on Build GRCh38
rs898456805 Chr.15: 72288418-72288418 on Build GRCh38
rs754228004 Chr.15: 72288464-72288464 on Build GRCh38
rs775868240 Chr.15: 72288462-72288463 on Build GRCh38
rs774435292 Chr.15: 72288436-72288436 on Build GRCh38
rs769219284 Chr.15: 72288560-72288566 on Build GRCh38
rs746499365 Chr.15: 72288477-72288477 on Build GRCh38
rs779619183 Chr.15: 72288609-72288609 on Build GRCh38
rs749077189 Chr.15: 72288616-72288616 on Build GRCh38
rs200682140 Chr.15: 72288500-72288500 on Build GRCh38
rs868098010 Chr.15: 72288454-72288455 on Build GRCh38
rs201565193 Chr.15: 72288474-72288474 on Build GRCh38
rs371011481 Chr.15: 72288491-72288491 on Build GRCh38
rs756417098 Chr.15: 72288448-72288448 on Build GRCh38
rs761876062 Chr.15: 72288528-72288528 on Build GRCh38
rs764015623 Chr.15: 72288435-72288435 on Build GRCh38
rs1028122137 Chr.15: 72288461-72288461 on Build GRCh38
rs753286721 Chr.15: 72288597-72288597 on Build GRCh38
rs775278294 Chr.15: 72288414-72288414 on Build GRCh38
rs945155007 Chr.15: 72288575-72288575 on Build GRCh38
rs765811380 Chr.15: 72288591-72288591 on Build GRCh38
rs749142792 Chr.15: 72288507-72288507 on Build GRCh38
rs775535163 Chr.15: 72288608-72288608 on Build GRCh38
rs776937831 Chr.15: 72288596-72288613 on Build GRCh38
rs369857833 Chr.15: 72288581-72288581 on Build GRCh38
rs768401912 Chr.15: 72288621-72288621 on Build GRCh38

View Full Product Details