Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151577658-151577905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160A1
Entrez Gene ID
729830
Gene Name
family with sequence similarity 160 member A1
Gene Aliases
-
UniGene
Hs.633810
Gene Chromosome Location
Chr.4: 151409143-151670503 on Build GRCh38
Transcripts
NM_001109977.1 XM_005263199.1 XM_011532220.1 XM_011532221.2 XM_011532222.2 XM_011532223.2 XM_011532224.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618481 c.389A>C p.H130P Chr.4: 151577733-151577733 on Build GRCh38
1427550 c.347T>C p.I116T Chr.4: 151577691-151577691 on Build GRCh38
3825352 c.387G>A p.S129S Chr.4: 151577731-151577731 on Build GRCh38
1738996 c.379A>G p.T127A Chr.4: 151577723-151577723 on Build GRCh38
1736523 c.483G>T p.K161N Chr.4: 151577827-151577827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907414867 Chr.4: 151577730-151577730 on Build GRCh38
rs1019532849 Chr.4: 151577836-151577836 on Build GRCh38
rs767408729 Chr.4: 151577707-151577707 on Build GRCh38
rs543303426 Chr.4: 151577793-151577793 on Build GRCh38
rs1028774652 Chr.4: 151577810-151577810 on Build GRCh38
rs182612729 Chr.4: 151577714-151577714 on Build GRCh38
rs747116506 Chr.4: 151577862-151577862 on Build GRCh38
rs969154788 Chr.4: 151577696-151577696 on Build GRCh38
rs200326017 Chr.4: 151577723-151577723 on Build GRCh38
rs555525555 Chr.4: 151577827-151577827 on Build GRCh38
rs753532630 Chr.4: 151577733-151577733 on Build GRCh38
rs768487408 Chr.4: 151577697-151577697 on Build GRCh38
rs1011656579 Chr.4: 151577801-151577801 on Build GRCh38
rs889790060 Chr.4: 151577812-151577812 on Build GRCh38
rs867037082 Chr.4: 151577843-151577843 on Build GRCh38
rs976908771 Chr.4: 151577791-151577791 on Build GRCh38
rs150755896 Chr.4: 151577874-151577874 on Build GRCh38
rs756800958 Chr.4: 151577754-151577754 on Build GRCh38
rs777352940 Chr.4: 151577731-151577731 on Build GRCh38
rs911464659 Chr.4: 151577739-151577739 on Build GRCh38
rs779441941 Chr.4: 151577759-151577759 on Build GRCh38
rs916242235 Chr.4: 151577686-151577686 on Build GRCh38
rs780685999 Chr.4: 151577814-151577814 on Build GRCh38
rs770845962 Chr.4: 151577734-151577734 on Build GRCh38
rs746193101 Chr.4: 151577788-151577788 on Build GRCh38

View Full Product Details