Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16969814-16970085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB4
Entrez Gene ID
128954
Gene Name
GRB2 associated binding protein family member 4
Gene Aliases
-
UniGene
Hs.434113
Gene Chromosome Location
Chr.22: 16961936-17008281 on Build GRCh38
Transcripts
NM_001037814.1 XM_011546114.2 XM_011546115.1 XM_011546116.2 XM_011546117.1 XM_017028575.1 XM_017028576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3405498 c.937+1G>A p.? Chr.22: 16969942-16969942 on Build GRCh38
5038051 c.866G>T p.R289I Chr.22: 16970014-16970014 on Build GRCh38
5898049 c.928G>A p.D310N Chr.22: 16969952-16969952 on Build GRCh38
1252924 c.877A>C p.S293R Chr.22: 16970003-16970003 on Build GRCh38
5644295 c.855C>A p.G285G Chr.22: 16970025-16970025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370292871 Chr.22: 16969954-16969954 on Build GRCh38
rs769082245 Chr.22: 16969942-16969942 on Build GRCh38
rs774656634 Chr.22: 16969944-16969944 on Build GRCh38
rs11703655 Chr.22: 16970062-16970062 on Build GRCh38
rs775796787 Chr.22: 16969922-16969922 on Build GRCh38
rs756284558 Chr.22: 16969953-16969953 on Build GRCh38
rs368140874 Chr.22: 16970014-16970014 on Build GRCh38
rs961708166 Chr.22: 16970064-16970064 on Build GRCh38
rs762452630 Chr.22: 16969948-16969948 on Build GRCh38
rs769322657 Chr.22: 16970010-16970010 on Build GRCh38
rs370660026 Chr.22: 16970037-16970037 on Build GRCh38
rs981823532 Chr.22: 16970040-16970040 on Build GRCh38
rs556305609 Chr.22: 16969879-16969879 on Build GRCh38
rs747363257 Chr.22: 16970028-16970028 on Build GRCh38
rs920440970 Chr.22: 16969949-16969949 on Build GRCh38
rs200505777 Chr.22: 16970009-16970009 on Build GRCh38
rs757272657 Chr.22: 16970007-16970007 on Build GRCh38
rs539055537 Chr.22: 16969840-16969840 on Build GRCh38
rs773888655 Chr.22: 16970043-16970043 on Build GRCh38
rs61741409 Chr.22: 16970039-16970039 on Build GRCh38
rs768902627 Chr.22: 16970019-16970019 on Build GRCh38
rs202215160 Chr.22: 16970008-16970008 on Build GRCh38
rs779627579 Chr.22: 16970012-16970012 on Build GRCh38
rs751446104 Chr.22: 16969996-16969996 on Build GRCh38
rs763259047 Chr.22: 16969935-16969935 on Build GRCh38
rs777628564 Chr.22: 16969980-16969980 on Build GRCh38
rs780817915 Chr.22: 16969904-16969904 on Build GRCh38
rs61738794 Chr.22: 16969875-16969875 on Build GRCh38
rs577446953 Chr.22: 16969872-16969872 on Build GRCh38
rs758208034 Chr.22: 16969976-16969976 on Build GRCh38
rs559238038 Chr.22: 16969863-16969863 on Build GRCh38
rs770278440 Chr.22: 16969901-16969901 on Build GRCh38
rs940416400 Chr.22: 16969909-16969909 on Build GRCh38
rs201405868 Chr.22: 16969951-16969951 on Build GRCh38
rs372681804 Chr.22: 16969911-16969911 on Build GRCh38
rs769480411 Chr.22: 16969920-16969920 on Build GRCh38
rs377466804 Chr.22: 16969973-16969973 on Build GRCh38
rs367789494 Chr.22: 16970056-16970056 on Build GRCh38
rs190990651 Chr.22: 16970030-16970030 on Build GRCh38
rs34530806 Chr.22: 16970013-16970014 on Build GRCh38
rs373491367 Chr.22: 16969970-16969970 on Build GRCh38
rs760789604 Chr.22: 16970053-16970053 on Build GRCh38

View Full Product Details