Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200901008-200901251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf106
Entrez Gene ID
55765
Gene Name
chromosome 1 open reading frame 106
Gene Aliases
-
UniGene
Hs.518997
Gene Chromosome Location
Chr.1: 200891074-200915736 on Build GRCh38
Transcripts
NM_001142569.2 NM_018265.3 XM_011509754.2 XM_011509755.1 XM_011509756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3785106 c.760C>T p.L254F Chr.1: 200901144-200901144 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888846117 Chr.1: 200901157-200901157 on Build GRCh38
rs935457798 Chr.1: 200901102-200901102 on Build GRCh38
rs751878605 Chr.1: 200901030-200901030 on Build GRCh38
rs138751093 Chr.1: 200901039-200901039 on Build GRCh38
rs780166208 Chr.1: 200901180-200901180 on Build GRCh38
rs750580083 Chr.1: 200901148-200901148 on Build GRCh38
rs769763675 Chr.1: 200901174-200901174 on Build GRCh38
rs982242559 Chr.1: 200901156-200901156 on Build GRCh38
rs751151214 Chr.1: 200901098-200901098 on Build GRCh38
rs917945569 Chr.1: 200901105-200901105 on Build GRCh38
rs774931721 Chr.1: 200901115-200901115 on Build GRCh38
rs543461914 Chr.1: 200901123-200901123 on Build GRCh38
rs1009025159 Chr.1: 200901230-200901230 on Build GRCh38
rs556785931 Chr.1: 200901144-200901144 on Build GRCh38
rs1050425909 Chr.1: 200901221-200901221 on Build GRCh38
rs369240943 Chr.1: 200901037-200901037 on Build GRCh38
rs897646225 Chr.1: 200901134-200901134 on Build GRCh38
rs924143423 Chr.1: 200901166-200901166 on Build GRCh38
rs997256958 Chr.1: 200901052-200901052 on Build GRCh38
rs536183830 Chr.1: 200901082-200901082 on Build GRCh38
rs756147177 Chr.1: 200901045-200901045 on Build GRCh38
rs879551515 Chr.1: 200901150-200901150 on Build GRCh38
rs956856211 Chr.1: 200901231-200901231 on Build GRCh38
rs757620702 Chr.1: 200901036-200901036 on Build GRCh38
rs948090808 Chr.1: 200901106-200901106 on Build GRCh38
rs931971607 Chr.1: 200901211-200901211 on Build GRCh38
rs377083257 Chr.1: 200901069-200901069 on Build GRCh38
rs750717551 Chr.1: 200901040-200901040 on Build GRCh38
rs780226969 Chr.1: 200901046-200901046 on Build GRCh38
rs952437333 Chr.1: 200901129-200901129 on Build GRCh38
rs759408185 Chr.1: 200901114-200901114 on Build GRCh38
rs376170884 Chr.1: 200901104-200901104 on Build GRCh38
rs752073272 Chr.1: 200901128-200901128 on Build GRCh38
rs1030028561 Chr.1: 200901053-200901053 on Build GRCh38
rs373530210 Chr.1: 200901179-200901179 on Build GRCh38
rs1036227894 Chr.1: 200901133-200901133 on Build GRCh38
rs935568758 Chr.1: 200901168-200901168 on Build GRCh38
rs777884844 Chr.1: 200901086-200901086 on Build GRCh38
rs767992876 Chr.1: 200901146-200901146 on Build GRCh38
rs970452466 Chr.1: 200901149-200901149 on Build GRCh38
rs887661614 Chr.1: 200901034-200901034 on Build GRCh38
rs141815054 Chr.1: 200901094-200901094 on Build GRCh38
rs890506998 Chr.1: 200901227-200901227 on Build GRCh38
rs770814611 Chr.1: 200901093-200901093 on Build GRCh38
rs961589415 Chr.1: 200901120-200901120 on Build GRCh38
rs762481350 Chr.1: 200901122-200901122 on Build GRCh38
rs972711786 Chr.1: 200901121-200901121 on Build GRCh38

View Full Product Details