Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14409224-14409479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX39A
Entrez Gene ID
10212
Gene Name
DEAD-box helicase 39A
Gene Aliases
BAT1, BAT1L, DDX39, DDXL, URH49
UniGene
Hs.311609
Gene Chromosome Location
Chr.19: 14408798-14419383 on Build GRCh38
Transcripts
NM_005804.3 NR_046366.2 XM_006722606.3 XM_011527620.1 XM_011527621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458848 c.1100C>T p.S367L Chr.19: 14409322-14409322 on Build GRCh38
4642261 c.989A>G p.Q330R Chr.19: 14409433-14409433 on Build GRCh38
7151710 c.980C>T p.S327L Chr.19: 14409442-14409442 on Build GRCh38
4676809 c.1009C>T p.R337W Chr.19: 14409413-14409413 on Build GRCh38
7184071 c.1071C>T p.I357I Chr.19: 14409351-14409351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73517823 Chr.19: 14409267-14409267 on Build GRCh38
rs550995811 Chr.19: 14409446-14409446 on Build GRCh38
rs897628666 Chr.19: 14409361-14409361 on Build GRCh38
rs992761315 Chr.19: 14409249-14409249 on Build GRCh38
rs751299237 Chr.19: 14409350-14409350 on Build GRCh38
rs769842789 Chr.19: 14409272-14409272 on Build GRCh38
rs771494825 Chr.19: 14409440-14409440 on Build GRCh38
rs138589818 Chr.19: 14409453-14409453 on Build GRCh38
rs1027698053 Chr.19: 14409387-14409387 on Build GRCh38
rs61406319 Chr.19: 14409375-14409375 on Build GRCh38
rs141070619 Chr.19: 14409351-14409351 on Build GRCh38
rs111233752 Chr.19: 14409449-14409449 on Build GRCh38
rs761572821 Chr.19: 14409454-14409454 on Build GRCh38
rs773575173 Chr.19: 14409438-14409438 on Build GRCh38
rs767211053 Chr.19: 14409459-14409459 on Build GRCh38
rs750184668 Chr.19: 14409366-14409366 on Build GRCh38
rs962179910 Chr.19: 14409383-14409386 on Build GRCh38
rs768663773 Chr.19: 14409391-14409391 on Build GRCh38
rs775652080 Chr.19: 14409284-14409284 on Build GRCh38
rs761187273 Chr.19: 14409305-14409305 on Build GRCh38
rs763639317 Chr.19: 14409343-14409343 on Build GRCh38
rs760992758 Chr.19: 14409439-14409439 on Build GRCh38
rs972288900 Chr.19: 14409399-14409399 on Build GRCh38
rs761998547 Chr.19: 14409293-14409293 on Build GRCh38
rs370950904 Chr.19: 14409266-14409266 on Build GRCh38
rs188717433 Chr.19: 14409412-14409412 on Build GRCh38
rs772264761 Chr.19: 14409426-14409426 on Build GRCh38
rs144815403 Chr.19: 14409355-14409355 on Build GRCh38
rs1050043686 Chr.19: 14409312-14409312 on Build GRCh38
rs756125583 Chr.19: 14409369-14409369 on Build GRCh38
rs746150070 Chr.19: 14409287-14409288 on Build GRCh38
rs1033992786 Chr.19: 14409342-14409342 on Build GRCh38
rs758178957 Chr.19: 14409393-14409393 on Build GRCh38
rs781680973 Chr.19: 14409255-14409255 on Build GRCh38
rs544169400 Chr.19: 14409252-14409252 on Build GRCh38
rs201814456 Chr.19: 14409339-14409339 on Build GRCh38
rs368220686 Chr.19: 14409268-14409268 on Build GRCh38
rs749406314 Chr.19: 14409378-14409378 on Build GRCh38
rs754652478 Chr.19: 14409294-14409294 on Build GRCh38
rs563136299 Chr.19: 14409451-14409451 on Build GRCh38
rs748432095 Chr.19: 14409413-14409413 on Build GRCh38
rs151091447 Chr.19: 14409321-14409321 on Build GRCh38
rs557071355 Chr.19: 14409297-14409297 on Build GRCh38
rs371904781 Chr.19: 14409288-14409288 on Build GRCh38
rs184015422 Chr.19: 14409289-14409289 on Build GRCh38
rs984466720 Chr.19: 14409374-14409374 on Build GRCh38
rs777056413 Chr.19: 14409444-14409444 on Build GRCh38
rs112129210 Chr.19: 14409259-14409259 on Build GRCh38

View Full Product Details