Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8459512-8459785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC6A
Entrez Gene ID
93978
Gene Name
C-type lectin domain family 6 member A
Gene Aliases
CLEC4N, CLECSF10
UniGene
Hs.591147
Gene Chromosome Location
Chr.12: 8455995-8478330 on Build GRCh38
Transcripts
NM_001007033.1 NM_001317999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
50381 c.203G>C p.S68T Chr.12: 8459678-8459678 on Build GRCh38
4918966 c.183T>C p.H61H Chr.12: 8459658-8459658 on Build GRCh38
405126 c.151delG p.G51fs*80 Chr.12: 8459626-8459626 on Build GRCh38
4412164 c.162G>C p.L54L Chr.12: 8459637-8459637 on Build GRCh38
1747277 c.145G>A p.E49K Chr.12: 8459620-8459620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938570101 Chr.12: 8459618-8459618 on Build GRCh38
rs747299245 Chr.12: 8459648-8459648 on Build GRCh38
rs779583761 Chr.12: 8459547-8459547 on Build GRCh38
rs757785611 Chr.12: 8459739-8459739 on Build GRCh38
rs747523136 Chr.12: 8459649-8459649 on Build GRCh38
rs749562245 Chr.12: 8459746-8459746 on Build GRCh38
rs760467769 Chr.12: 8459697-8459697 on Build GRCh38
rs200251402 Chr.12: 8459563-8459563 on Build GRCh38
rs778349953 Chr.12: 8459644-8459644 on Build GRCh38
rs371035585 Chr.12: 8459685-8459685 on Build GRCh38
rs763125854 Chr.12: 8459549-8459549 on Build GRCh38
rs10770736 Chr.12: 8459740-8459740 on Build GRCh38
rs947259080 Chr.12: 8459547-8459547 on Build GRCh38
rs756792941 Chr.12: 8459626-8459626 on Build GRCh38
rs774456308 Chr.12: 8459683-8459683 on Build GRCh38
rs753514994 Chr.12: 8459705-8459705 on Build GRCh38
rs754682432 Chr.12: 8459698-8459708 on Build GRCh38
rs773508357 Chr.12: 8459548-8459548 on Build GRCh38
rs781464279 Chr.12: 8459654-8459654 on Build GRCh38
rs770212139 Chr.12: 8459667-8459667 on Build GRCh38
rs966853045 Chr.12: 8459613-8459613 on Build GRCh38
rs138045958 Chr.12: 8459625-8459625 on Build GRCh38
rs532208747 Chr.12: 8459741-8459741 on Build GRCh38
rs777418173 Chr.12: 8459606-8459606 on Build GRCh38
rs763216958 Chr.12: 8459673-8459673 on Build GRCh38
rs771183062 Chr.12: 8459678-8459678 on Build GRCh38
rs142509792 Chr.12: 8459658-8459658 on Build GRCh38
rs750615757 Chr.12: 8459637-8459637 on Build GRCh38
rs752530953 Chr.12: 8459584-8459584 on Build GRCh38
rs769178216 Chr.12: 8459651-8459651 on Build GRCh38
rs773596201 Chr.12: 8459670-8459670 on Build GRCh38
rs796098130 Chr.12: 8459629-8459629 on Build GRCh38
rs759495670 Chr.12: 8459566-8459566 on Build GRCh38
rs778329997 Chr.12: 8459709-8459709 on Build GRCh38
rs766523123 Chr.12: 8459552-8459552 on Build GRCh38
rs755782205 Chr.12: 8459588-8459588 on Build GRCh38
rs749948176 Chr.12: 8459717-8459717 on Build GRCh38
rs752520350 Chr.12: 8459693-8459693 on Build GRCh38
rs150852294 Chr.12: 8459698-8459698 on Build GRCh38
rs1041511458 Chr.12: 8459579-8459579 on Build GRCh38
rs766171537 Chr.12: 8459579-8459581 on Build GRCh38
rs374604334 Chr.12: 8459689-8459689 on Build GRCh38
rs749837822 Chr.12: 8459730-8459730 on Build GRCh38
rs767254621 Chr.12: 8459567-8459567 on Build GRCh38

View Full Product Details