Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58645556-58645781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3D
Entrez Gene ID
131177
Gene Name
family with sequence similarity 3 member D
Gene Aliases
EF7, OIT1
UniGene
Hs.61265
Gene Chromosome Location
Chr.3: 58633943-58666843 on Build GRCh38
Transcripts
NM_138805.2 XM_005264862.4 XM_005264863.4 XM_005264864.4 XM_006712965.3 XM_006712967.3 XM_011533348.2 XM_011533349.2 XM_011533350.2 XM_011533351.2 XM_011533352.2 XM_011533353.2 XM_011533354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1309418 c.146-1G>C p.? Chr.3: 58645627-58645627 on Build GRCh38
7209662 c.183C>T p.C61C Chr.3: 58645589-58645589 on Build GRCh38
6006973 c.188C>T p.A63V Chr.3: 58645584-58645584 on Build GRCh38
7178743 c.171C>T p.L57L Chr.3: 58645601-58645601 on Build GRCh38
5865178 c.184C>T p.P62S Chr.3: 58645588-58645588 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769339261 Chr.3: 58645592-58645592 on Build GRCh38
rs200028657 Chr.3: 58645693-58645704 on Build GRCh38
rs143089006 Chr.3: 58645715-58645715 on Build GRCh38
rs573206608 Chr.3: 58645655-58645655 on Build GRCh38
rs372504404 Chr.3: 58645748-58645748 on Build GRCh38
rs763950619 Chr.3: 58645636-58645636 on Build GRCh38
rs147876713 Chr.3: 58645607-58645607 on Build GRCh38
rs764917440 Chr.3: 58645676-58645676 on Build GRCh38
rs370131941 Chr.3: 58645616-58645616 on Build GRCh38
rs535746548 Chr.3: 58645668-58645668 on Build GRCh38
rs375479665 Chr.3: 58645759-58645759 on Build GRCh38
rs747547903 Chr.3: 58645585-58645585 on Build GRCh38
rs17059569 Chr.3: 58645594-58645594 on Build GRCh38
rs770054191 Chr.3: 58645603-58645603 on Build GRCh38
rs756988687 Chr.3: 58645678-58645688 on Build GRCh38
rs146148015 Chr.3: 58645740-58645740 on Build GRCh38
rs555583943 Chr.3: 58645696-58645696 on Build GRCh38
rs994146687 Chr.3: 58645739-58645739 on Build GRCh38
rs865911193 Chr.3: 58645716-58645716 on Build GRCh38
rs762838006 Chr.3: 58645755-58645755 on Build GRCh38
rs952748179 Chr.3: 58645747-58645747 on Build GRCh38
rs757238293 Chr.3: 58645587-58645587 on Build GRCh38
rs773094560 Chr.3: 58645618-58645618 on Build GRCh38
rs1044848925 Chr.3: 58645687-58645687 on Build GRCh38
rs540275482 Chr.3: 58645760-58645760 on Build GRCh38
rs374555282 Chr.3: 58645598-58645598 on Build GRCh38
rs904209347 Chr.3: 58645597-58645597 on Build GRCh38
rs1031393697 Chr.3: 58645602-58645602 on Build GRCh38
rs373820724 Chr.3: 58645674-58645674 on Build GRCh38
rs753878891 Chr.3: 58645676-58645677 on Build GRCh38
rs752433761 Chr.3: 58645678-58645678 on Build GRCh38
rs553311501 Chr.3: 58645610-58645610 on Build GRCh38
rs895796025 Chr.3: 58645719-58645719 on Build GRCh38
rs867687354 Chr.3: 58645723-58645723 on Build GRCh38
rs140725039 Chr.3: 58645584-58645584 on Build GRCh38
rs755636267 Chr.3: 58645624-58645626 on Build GRCh38
rs759025588 Chr.3: 58645675-58645675 on Build GRCh38
rs187056895 Chr.3: 58645593-58645593 on Build GRCh38

View Full Product Details