Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49732506-49732765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM117A
Entrez Gene ID
81558
Gene Name
family with sequence similarity 117 member A
Gene Aliases
-
UniGene
Hs.514308
Gene Chromosome Location
Chr.17: 49710325-49764156 on Build GRCh38
Transcripts
NM_030802.3 XM_017025182.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
980836 c.213G>A p.S71S Chr.17: 49732704-49732704 on Build GRCh38
4483844 c.273C>T p.S91S Chr.17: 49732644-49732644 on Build GRCh38
1251711 c.259C>T p.R87W Chr.17: 49732658-49732658 on Build GRCh38
5881356 c.325G>T p.D109Y Chr.17: 49732592-49732592 on Build GRCh38
5386632 c.305G>A p.G102D Chr.17: 49732612-49732612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761925203 Chr.17: 49732588-49732588 on Build GRCh38
rs527239384 Chr.17: 49732740-49732740 on Build GRCh38
rs765436383 Chr.17: 49732595-49732595 on Build GRCh38
rs202016558 Chr.17: 49732657-49732657 on Build GRCh38
rs755529128 Chr.17: 49732655-49732655 on Build GRCh38
rs774214940 Chr.17: 49732530-49732530 on Build GRCh38
rs540981062 Chr.17: 49732703-49732703 on Build GRCh38
rs771671869 Chr.17: 49732629-49732629 on Build GRCh38
rs765311423 Chr.17: 49732708-49732708 on Build GRCh38
rs372734158 Chr.17: 49732541-49732541 on Build GRCh38
rs1044647471 Chr.17: 49732668-49732668 on Build GRCh38
rs768610206 Chr.17: 49732702-49732702 on Build GRCh38
rs773352811 Chr.17: 49732710-49732710 on Build GRCh38
rs570058397 Chr.17: 49732739-49732739 on Build GRCh38
rs761863833 Chr.17: 49732704-49732704 on Build GRCh38
rs750698367 Chr.17: 49732600-49732600 on Build GRCh38
rs141538086 Chr.17: 49732654-49732654 on Build GRCh38
rs764034170 Chr.17: 49732560-49732560 on Build GRCh38
rs745646169 Chr.17: 49732538-49732538 on Build GRCh38
rs775380216 Chr.17: 49732686-49732686 on Build GRCh38
rs778599058 Chr.17: 49732745-49732745 on Build GRCh38
rs187054709 Chr.17: 49732573-49732573 on Build GRCh38
rs760701479 Chr.17: 49732546-49732546 on Build GRCh38
rs993734010 Chr.17: 49732718-49732718 on Build GRCh38
rs766601875 Chr.17: 49732729-49732729 on Build GRCh38
rs746832118 Chr.17: 49732699-49732699 on Build GRCh38
rs191416134 Chr.17: 49732743-49732743 on Build GRCh38
rs745521538 Chr.17: 49732684-49732684 on Build GRCh38
rs552721803 Chr.17: 49732744-49732744 on Build GRCh38
rs373170061 Chr.17: 49732715-49732715 on Build GRCh38
rs377253020 Chr.17: 49732647-49732647 on Build GRCh38
rs909379986 Chr.17: 49732584-49732584 on Build GRCh38
rs748776433 Chr.17: 49732659-49732659 on Build GRCh38
rs771955381 Chr.17: 49732685-49732685 on Build GRCh38
rs370182807 Chr.17: 49732681-49732681 on Build GRCh38
rs888799040 Chr.17: 49732583-49732583 on Build GRCh38
rs1007223063 Chr.17: 49732594-49732594 on Build GRCh38
rs11539791 Chr.17: 49732606-49732606 on Build GRCh38
rs756886045 Chr.17: 49732661-49732661 on Build GRCh38
rs768055906 Chr.17: 49732742-49732742 on Build GRCh38
rs766767954 Chr.17: 49732649-49732649 on Build GRCh38
rs763191016 Chr.17: 49732716-49732716 on Build GRCh38
rs751930234 Chr.17: 49732734-49732734 on Build GRCh38

View Full Product Details