Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154332593-154332818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B2
Entrez Gene ID
57198
Gene Name
ATPase phospholipid transporting 8B2
Gene Aliases
ATPID
UniGene
Hs.435700
Gene Chromosome Location
Chr.1: 154325541-154351307 on Build GRCh38
Transcripts
NM_001005855.1 NM_020452.3 XM_005245355.2 XM_005245356.3 XM_017001871.1 XM_017001872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5453299 c.588C>T p.D196D Chr.1: 154332696-154332696 on Build GRCh38
5453300 c.645C>T p.D215D Chr.1: 154332696-154332696 on Build GRCh38
5159892 c.646A>G p.T216A Chr.1: 154332655-154332655 on Build GRCh38
7147405 c.688G>A p.G230S Chr.1: 154332697-154332697 on Build GRCh38
6345266 c.661G>T p.D221Y Chr.1: 154332670-154332670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887276399 Chr.1: 154332780-154332780 on Build GRCh38
rs996006427 Chr.1: 154332736-154332736 on Build GRCh38
rs1047205115 Chr.1: 154332758-154332758 on Build GRCh38
rs377214445 Chr.1: 154332723-154332723 on Build GRCh38
rs1008399775 Chr.1: 154332792-154332792 on Build GRCh38
rs778919135 Chr.1: 154332664-154332664 on Build GRCh38
rs12077870 Chr.1: 154332714-154332714 on Build GRCh38
rs756497833 Chr.1: 154332648-154332648 on Build GRCh38
rs981632475 Chr.1: 154332638-154332638 on Build GRCh38
rs759045478 Chr.1: 154332718-154332718 on Build GRCh38
rs371131370 Chr.1: 154332666-154332666 on Build GRCh38
rs759970744 Chr.1: 154332626-154332626 on Build GRCh38
rs112957714 Chr.1: 154332778-154332778 on Build GRCh38
rs769630285 Chr.1: 154332697-154332697 on Build GRCh38
rs150253336 Chr.1: 154332772-154332772 on Build GRCh38
rs748947635 Chr.1: 154332711-154332711 on Build GRCh38
rs1049841745 Chr.1: 154332738-154332738 on Build GRCh38
rs754154734 Chr.1: 154332653-154332653 on Build GRCh38
rs577186673 Chr.1: 154332696-154332696 on Build GRCh38
rs776822568 Chr.1: 154332760-154332760 on Build GRCh38
rs937363337 Chr.1: 154332656-154332656 on Build GRCh38
rs141051422 Chr.1: 154332645-154332645 on Build GRCh38
rs781069879 Chr.1: 154332669-154332669 on Build GRCh38
rs561991401 Chr.1: 154332713-154332713 on Build GRCh38
rs745904972 Chr.1: 154332665-154332665 on Build GRCh38
rs757497332 Chr.1: 154332658-154332658 on Build GRCh38
rs778259766 Chr.1: 154332652-154332652 on Build GRCh38
rs774283149 Chr.1: 154332744-154332744 on Build GRCh38
rs576546996 Chr.1: 154332682-154332682 on Build GRCh38
rs562571797 Chr.1: 154332759-154332759 on Build GRCh38
rs776187208 Chr.1: 154332719-154332719 on Build GRCh38
rs770556905 Chr.1: 154332713-154332713 on Build GRCh38
rs1045687634 Chr.1: 154332618-154332618 on Build GRCh38
rs772983983 Chr.1: 154332705-154332705 on Build GRCh38
rs768000646 Chr.1: 154332637-154332637 on Build GRCh38

View Full Product Details