Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93479007-93479232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO4
Entrez Gene ID
56935
Gene Name
single-pass membrane protein with coiled-coil domains 4
Gene Aliases
C11orf75, FN5
UniGene
Hs.438064
Gene Chromosome Location
Chr.11: 93478472-93586856 on Build GRCh38
Transcripts
NM_020179.2 XM_011542907.1 XM_011542908.2 XM_011542909.1 XM_011542910.2 XM_011542911.2 XM_017018019.1 XM_017018020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5717616 c.135C>T p.I45I Chr.11: 93479055-93479055 on Build GRCh38
5789518 c.5G>C p.R2P Chr.11: 93479185-93479185 on Build GRCh38
5051211 c.111G>A p.T37T Chr.11: 93479079-93479079 on Build GRCh38
5619307 c.102G>A p.V34V Chr.11: 93479088-93479088 on Build GRCh38
3454689 c.12C>T p.L4L Chr.11: 93479178-93479178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764757111 Chr.11: 93479080-93479080 on Build GRCh38
rs760940483 Chr.11: 93479201-93479201 on Build GRCh38
rs370993774 Chr.11: 93479094-93479094 on Build GRCh38
rs745421729 Chr.11: 93479162-93479162 on Build GRCh38
rs755033880 Chr.11: 93479130-93479130 on Build GRCh38
rs763585208 Chr.11: 93479213-93479214 on Build GRCh38
rs972412916 Chr.11: 93479055-93479055 on Build GRCh38
rs141230765 Chr.11: 93479129-93479129 on Build GRCh38
rs202058974 Chr.11: 93479030-93479030 on Build GRCh38
rs762437359 Chr.11: 93479186-93479186 on Build GRCh38
rs770207119 Chr.11: 93479066-93479075 on Build GRCh38
rs528994325 Chr.11: 93479067-93479067 on Build GRCh38
rs780202294 Chr.11: 93479040-93479040 on Build GRCh38
rs767850204 Chr.11: 93479189-93479189 on Build GRCh38
rs2658797 Chr.11: 93479088-93479088 on Build GRCh38
rs754036634 Chr.11: 93479204-93479204 on Build GRCh38
rs757684648 Chr.11: 93479029-93479029 on Build GRCh38
rs140592708 Chr.11: 93479135-93479135 on Build GRCh38
rs770433714 Chr.11: 93479072-93479072 on Build GRCh38
rs569882632 Chr.11: 93479185-93479185 on Build GRCh38
rs142188395 Chr.11: 93479061-93479061 on Build GRCh38
rs750629862 Chr.11: 93479032-93479032 on Build GRCh38
rs746715784 Chr.11: 93479142-93479142 on Build GRCh38
rs771831111 Chr.11: 93479064-93479064 on Build GRCh38
rs549244127 Chr.11: 93479213-93479213 on Build GRCh38
rs893001312 Chr.11: 93479051-93479051 on Build GRCh38
rs756369348 Chr.11: 93479110-93479110 on Build GRCh38
rs892694781 Chr.11: 93479188-93479188 on Build GRCh38
rs909072912 Chr.11: 93479039-93479039 on Build GRCh38
rs749264689 Chr.11: 93479044-93479044 on Build GRCh38
rs904851628 Chr.11: 93479211-93479211 on Build GRCh38
rs756253689 Chr.11: 93479034-93479034 on Build GRCh38
rs371845014 Chr.11: 93479182-93479182 on Build GRCh38
rs537227026 Chr.11: 93479209-93479209 on Build GRCh38
rs773626137 Chr.11: 93479198-93479198 on Build GRCh38
rs766439976 Chr.11: 93479111-93479111 on Build GRCh38
rs776136865 Chr.11: 93479148-93479148 on Build GRCh38
rs1035063441 Chr.11: 93479102-93479102 on Build GRCh38
rs778869372 Chr.11: 93479132-93479132 on Build GRCh38
rs777645014 Chr.11: 93479141-93479141 on Build GRCh38
rs138064563 Chr.11: 93479024-93479024 on Build GRCh38
rs752094282 Chr.11: 93479083-93479083 on Build GRCh38
rs3207348 Chr.11: 93479079-93479079 on Build GRCh38
rs752037332 Chr.11: 93479028-93479028 on Build GRCh38
rs199696547 Chr.11: 93479066-93479066 on Build GRCh38
rs770683740 Chr.11: 93479146-93479146 on Build GRCh38
rs748044004 Chr.11: 93479134-93479134 on Build GRCh38
rs773804914 Chr.11: 93479191-93479191 on Build GRCh38
rs776210014 Chr.11: 93479073-93479073 on Build GRCh38
rs542887920 Chr.11: 93479052-93479052 on Build GRCh38
rs1039785592 Chr.11: 93479175-93479175 on Build GRCh38
rs779314812 Chr.11: 93479203-93479203 on Build GRCh38

View Full Product Details