Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108210123-108210368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG4
Entrez Gene ID
3981
Gene Name
DNA ligase 4
Gene Aliases
LIG4S
UniGene
Hs.166091
Gene Chromosome Location
Chr.13: 108207442-108218368 on Build GRCh38
Transcripts
NM_001098268.1 NM_002312.3 NM_206937.1 XM_005254056.1 XM_005254057.4 XM_005254058.3 XM_006719951.3 XM_006719952.1 XM_011521091.2 XM_011521092.2 XM_017020563.1 XM_017020564.1 XM_017020565.1 XM_017020566.1 XM_017020567.1 XM_017020568.1 XM_017020569.1 XM_017020570.1 XM_017020571.1 XM_017020572.1 XM_017020573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6432414 c.1145T>A p.L382Q Chr.13: 108210124-108210124 on Build GRCh38
7288801 c.960A>G p.K320K Chr.13: 108210309-108210309 on Build GRCh38
4876607 c.1135C>T p.H379Y Chr.13: 108210134-108210134 on Build GRCh38
696071 c.1067A>T p.E356V Chr.13: 108210202-108210202 on Build GRCh38
3704508 c.973A>G p.I325V Chr.13: 108210296-108210296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369400986 Chr.13: 108210334-108210334 on Build GRCh38
rs549389506 Chr.13: 108210253-108210253 on Build GRCh38
rs774604168 Chr.13: 108210265-108210265 on Build GRCh38
rs757762017 Chr.13: 108210310-108210310 on Build GRCh38
rs767610700 Chr.13: 108210300-108210300 on Build GRCh38
rs756549415 Chr.13: 108210162-108210162 on Build GRCh38
rs764497174 Chr.13: 108210304-108210304 on Build GRCh38
rs746602134 Chr.13: 108210201-108210201 on Build GRCh38
rs754406950 Chr.13: 108210177-108210177 on Build GRCh38
rs144322912 Chr.13: 108210175-108210175 on Build GRCh38
rs760940573 Chr.13: 108210302-108210302 on Build GRCh38
rs747868033 Chr.13: 108210221-108210221 on Build GRCh38
rs767884120 Chr.13: 108210156-108210156 on Build GRCh38
rs535025599 Chr.13: 108210249-108210249 on Build GRCh38
rs780950741 Chr.13: 108210208-108210208 on Build GRCh38
rs754282095 Chr.13: 108210308-108210308 on Build GRCh38
rs371477700 Chr.13: 108210186-108210186 on Build GRCh38
rs753025101 Chr.13: 108210158-108210158 on Build GRCh38
rs769699485 Chr.13: 108210237-108210237 on Build GRCh38
rs146091097 Chr.13: 108210199-108210199 on Build GRCh38
rs376108062 Chr.13: 108210301-108210301 on Build GRCh38
rs759754809 Chr.13: 108210269-108210269 on Build GRCh38
rs566324623 Chr.13: 108210200-108210200 on Build GRCh38
rs750391445 Chr.13: 108210174-108210179 on Build GRCh38
rs762741359 Chr.13: 108210335-108210336 on Build GRCh38
rs754614183 Chr.13: 108210204-108210204 on Build GRCh38
rs773271264 Chr.13: 108210244-108210244 on Build GRCh38

View Full Product Details