Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34726734-34726977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM205A
Entrez Gene ID
259308
Gene Name
family with sequence similarity 205 member A
Gene Aliases
C9orf144B
UniGene
Hs.522093
Gene Chromosome Location
Chr.9: 34723053-34729538 on Build GRCh38
Transcripts
NM_001141917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7449380 c.363G>C p.E121D Chr.9: 34726877-34726877 on Build GRCh38
4839334 c.431C>T p.T144M Chr.9: 34726809-34726809 on Build GRCh38
5642871 c.329T>C p.V110A Chr.9: 34726911-34726911 on Build GRCh38
4187024 c.414C>A p.C138* Chr.9: 34726826-34726826 on Build GRCh38
5576645 c.388T>A p.S130T Chr.9: 34726852-34726852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141945649 Chr.9: 34726954-34726954 on Build GRCh38
rs564963123 Chr.9: 34726957-34726957 on Build GRCh38
rs751344372 Chr.9: 34726804-34726804 on Build GRCh38
rs1028369832 Chr.9: 34726763-34726763 on Build GRCh38
rs753053164 Chr.9: 34726939-34726939 on Build GRCh38
rs892890967 Chr.9: 34726944-34726944 on Build GRCh38
rs1055134568 Chr.9: 34726818-34726818 on Build GRCh38
rs936850415 Chr.9: 34726865-34726865 on Build GRCh38
rs757141318 Chr.9: 34726805-34726805 on Build GRCh38
rs193234006 Chr.9: 34726851-34726851 on Build GRCh38
rs961177405 Chr.9: 34726802-34726802 on Build GRCh38
rs755841009 Chr.9: 34726857-34726857 on Build GRCh38
rs758718497 Chr.9: 34726947-34726947 on Build GRCh38
rs188872830 Chr.9: 34726850-34726850 on Build GRCh38
rs117956208 Chr.9: 34726824-34726824 on Build GRCh38
rs182687245 Chr.9: 34726784-34726784 on Build GRCh38
rs779416495 Chr.9: 34726923-34726923 on Build GRCh38
rs908414247 Chr.9: 34726797-34726797 on Build GRCh38
rs559668831 Chr.9: 34726953-34726953 on Build GRCh38
rs766846222 Chr.9: 34726847-34726847 on Build GRCh38
rs550998564 Chr.9: 34726956-34726956 on Build GRCh38
rs1054331599 Chr.9: 34726877-34726877 on Build GRCh38
rs775195387 Chr.9: 34726776-34726776 on Build GRCh38
rs938059657 Chr.9: 34726808-34726808 on Build GRCh38
rs190805976 Chr.9: 34726778-34726778 on Build GRCh38
rs949626988 Chr.9: 34726841-34726841 on Build GRCh38
rs868534729 Chr.9: 34726799-34726799 on Build GRCh38
rs779341341 Chr.9: 34726936-34726936 on Build GRCh38

View Full Product Details