Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165754871-165755108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT3
Entrez Gene ID
2591
Gene Name
polypeptide N-acetylgalactosaminyltransferase 3
Gene Aliases
GalNAc-T3, HFTC, HHS
UniGene
Hs.170986
Gene Chromosome Location
Chr.2: 165747803-165796352 on Build GRCh38
Transcripts
NM_004482.3 XM_005246449.1 XM_011510929.1 XM_017003770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1482127 c.1441C>T p.Q481* Chr.2: 165755015-165755015 on Build GRCh38
340622 c.1428A>G p.I476M Chr.2: 165755028-165755028 on Build GRCh38
1400726 c.1486G>T p.V496L Chr.2: 165754970-165754970 on Build GRCh38
1008674 c.1524C>T p.Y508Y Chr.2: 165754932-165754932 on Build GRCh38
4627936 c.1515A>G p.I505M Chr.2: 165754941-165754941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753385900 Chr.2: 165755029-165755029 on Build GRCh38
rs775461827 Chr.2: 165754938-165754938 on Build GRCh38
rs753199090 Chr.2: 165755021-165755021 on Build GRCh38
rs575869340 Chr.2: 165755004-165755004 on Build GRCh38
rs757020982 Chr.2: 165755032-165755032 on Build GRCh38
rs759912211 Chr.2: 165755019-165755019 on Build GRCh38
rs747649996 Chr.2: 165754914-165754914 on Build GRCh38
rs1014664640 Chr.2: 165754939-165754939 on Build GRCh38
rs763224582 Chr.2: 165754982-165754982 on Build GRCh38
rs200667509 Chr.2: 165754923-165754923 on Build GRCh38
rs886055014 Chr.2: 165755005-165755005 on Build GRCh38
rs751916901 Chr.2: 165755008-165755008 on Build GRCh38
rs947920050 Chr.2: 165755052-165755052 on Build GRCh38
rs372619915 Chr.2: 165754917-165754917 on Build GRCh38
rs544166506 Chr.2: 165755075-165755075 on Build GRCh38
rs777492317 Chr.2: 165755025-165755025 on Build GRCh38
rs550354639 Chr.2: 165754920-165754920 on Build GRCh38
rs934481599 Chr.2: 165754950-165754950 on Build GRCh38
rs766572413 Chr.2: 165754996-165754996 on Build GRCh38
rs979264172 Chr.2: 165755058-165755058 on Build GRCh38
rs947021038 Chr.2: 165755059-165755059 on Build GRCh38
rs767939917 Chr.2: 165755020-165755020 on Build GRCh38
rs573424079 Chr.2: 165754893-165754893 on Build GRCh38
rs761823954 Chr.2: 165754943-165754943 on Build GRCh38
rs755699017 Chr.2: 165755022-165755022 on Build GRCh38
rs745655924 Chr.2: 165754931-165754931 on Build GRCh38
rs769810017 Chr.2: 165754961-165754961 on Build GRCh38
rs375879489 Chr.2: 165754927-165754927 on Build GRCh38
rs778598587 Chr.2: 165755042-165755042 on Build GRCh38
rs137853089 Chr.2: 165755015-165755015 on Build GRCh38
rs773423949 Chr.2: 165754970-165754970 on Build GRCh38
rs748891012 Chr.2: 165754921-165754921 on Build GRCh38
rs144331248 Chr.2: 165754932-165754932 on Build GRCh38
rs374205386 Chr.2: 165754975-165754975 on Build GRCh38

View Full Product Details