Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74187456-74187701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4H
Entrez Gene ID
7458
Gene Name
eukaryotic translation initiation factor 4H
Gene Aliases
WBSCR1, WSCR1, eIF-4H
UniGene
Hs.520943
Gene Chromosome Location
Chr.7: 74174376-74197101 on Build GRCh38
Transcripts
NM_022170.1 NM_031992.1 XM_011516556.1 XM_011516557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062565 c.149C>A p.P50H Chr.7: 74187700-74187700 on Build GRCh38
7108631 c.150T>G p.P50P Chr.7: 74187701-74187701 on Build GRCh38
3087543 c.67G>A p.G23S Chr.7: 74187618-74187618 on Build GRCh38
6337359 c.137T>A p.V46E Chr.7: 74187688-74187688 on Build GRCh38
6653063 c.92G>T p.R31L Chr.7: 74187643-74187643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138932705 Chr.7: 74187671-74187671 on Build GRCh38
rs987416273 Chr.7: 74187601-74187601 on Build GRCh38
rs371654694 Chr.7: 74187581-74187581 on Build GRCh38
rs1032560063 Chr.7: 74187527-74187527 on Build GRCh38
rs782781045 Chr.7: 74187520-74187520 on Build GRCh38
rs201237184 Chr.7: 74187668-74187668 on Build GRCh38
rs781910061 Chr.7: 74187573-74187573 on Build GRCh38
rs200405095 Chr.7: 74187617-74187617 on Build GRCh38
rs782551563 Chr.7: 74187620-74187620 on Build GRCh38
rs781855084 Chr.7: 74187602-74187602 on Build GRCh38
rs904920829 Chr.7: 74187522-74187522 on Build GRCh38
rs954660113 Chr.7: 74187543-74187543 on Build GRCh38
rs534331725 Chr.7: 74187531-74187531 on Build GRCh38
rs893647929 Chr.7: 74187530-74187530 on Build GRCh38
rs782793822 Chr.7: 74187590-74187590 on Build GRCh38
rs1001976016 Chr.7: 74187526-74187526 on Build GRCh38
rs782706514 Chr.7: 74187572-74187572 on Build GRCh38
rs965129504 Chr.7: 74187629-74187629 on Build GRCh38
rs920723246 Chr.7: 74187674-74187674 on Build GRCh38
rs782259889 Chr.7: 74187642-74187642 on Build GRCh38
rs782362109 Chr.7: 74187659-74187659 on Build GRCh38
rs782661706 Chr.7: 74187622-74187622 on Build GRCh38
rs111675936 Chr.7: 74187521-74187521 on Build GRCh38
rs184690257 Chr.7: 74187514-74187514 on Build GRCh38
rs1048013163 Chr.7: 74187484-74187484 on Build GRCh38
rs17146089 Chr.7: 74187536-74187536 on Build GRCh38
rs35523655 Chr.7: 74187611-74187611 on Build GRCh38
rs782088509 Chr.7: 74187481-74187481 on Build GRCh38
rs141344014 Chr.7: 74187618-74187618 on Build GRCh38
rs782148274 Chr.7: 74187582-74187582 on Build GRCh38
rs782479643 Chr.7: 74187607-74187607 on Build GRCh38
rs782619611 Chr.7: 74187660-74187660 on Build GRCh38
rs145187247 Chr.7: 74187662-74187662 on Build GRCh38
rs116438433 Chr.7: 74187539-74187539 on Build GRCh38
rs372807171 Chr.7: 74187542-74187542 on Build GRCh38

View Full Product Details