Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41436950-41437223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MX1
Entrez Gene ID
4599
Gene Name
MX dynamin like GTPase 1
Gene Aliases
IFI-78K, IFI78, MX, MxA
UniGene
Hs.517307
Gene Chromosome Location
Chr.21: 41420329-41459214 on Build GRCh38
Transcripts
NM_001144925.2 NM_001178046.2 NM_001282920.1 NM_002462.4 XM_005260978.4 XM_005260979.2 XM_005260980.2 XM_005260981.2 XM_005260982.2 XM_011529568.2 XM_011529570.2 XM_017028349.1 XM_017028350.1 XM_017028351.1 XM_017028352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5974237 c.392T>C p.I131T Chr.21: 41437108-41437108 on Build GRCh38
6161272 c.327G>A p.L109L Chr.21: 41437043-41437043 on Build GRCh38
6895985 c.316C>T p.P106S Chr.21: 41437032-41437032 on Build GRCh38
3550938 c.303C>T p.I101I Chr.21: 41437019-41437019 on Build GRCh38
4648451 c.319C>T p.L107L Chr.21: 41437035-41437035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767182695 Chr.21: 41436998-41436998 on Build GRCh38
rs752339991 Chr.21: 41437003-41437003 on Build GRCh38
rs774173358 Chr.21: 41437037-41437037 on Build GRCh38
rs763121684 Chr.21: 41436987-41436990 on Build GRCh38
rs768925279 Chr.21: 41436984-41436984 on Build GRCh38
rs749472574 Chr.21: 41437108-41437108 on Build GRCh38
rs768072252 Chr.21: 41437197-41437197 on Build GRCh38
rs767252754 Chr.21: 41437047-41437047 on Build GRCh38
rs550808214 Chr.21: 41437018-41437018 on Build GRCh38
rs761415634 Chr.21: 41437179-41437179 on Build GRCh38
rs768897139 Chr.21: 41437149-41437149 on Build GRCh38
rs377043349 Chr.21: 41437065-41437065 on Build GRCh38
rs763822126 Chr.21: 41437148-41437148 on Build GRCh38
rs907372877 Chr.21: 41437048-41437048 on Build GRCh38
rs546841301 Chr.21: 41437103-41437103 on Build GRCh38
rs557662342 Chr.21: 41437069-41437069 on Build GRCh38
rs1030165349 Chr.21: 41437178-41437178 on Build GRCh38
rs749928029 Chr.21: 41437190-41437190 on Build GRCh38
rs749382504 Chr.21: 41437020-41437020 on Build GRCh38
rs376506709 Chr.21: 41437111-41437111 on Build GRCh38
rs748539949 Chr.21: 41437100-41437100 on Build GRCh38
rs778153587 Chr.21: 41437106-41437106 on Build GRCh38
rs771162348 Chr.21: 41437109-41437109 on Build GRCh38
rs993382023 Chr.21: 41437177-41437177 on Build GRCh38
rs774454492 Chr.21: 41437110-41437110 on Build GRCh38
rs754222844 Chr.21: 41437163-41437163 on Build GRCh38
rs759473507 Chr.21: 41437046-41437046 on Build GRCh38
rs531283923 Chr.21: 41436978-41436978 on Build GRCh38
rs142837809 Chr.21: 41437086-41437086 on Build GRCh38
rs575820040 Chr.21: 41437127-41437127 on Build GRCh38
rs776288123 Chr.21: 41437159-41437159 on Build GRCh38
rs752039545 Chr.21: 41437085-41437085 on Build GRCh38
rs772030692 Chr.21: 41437112-41437112 on Build GRCh38
rs201692013 Chr.21: 41437098-41437098 on Build GRCh38
rs906108024 Chr.21: 41437033-41437033 on Build GRCh38
rs139708669 Chr.21: 41437034-41437034 on Build GRCh38
rs778070230 Chr.21: 41437146-41437146 on Build GRCh38
rs773259944 Chr.21: 41437164-41437165 on Build GRCh38
rs375569357 Chr.21: 41437117-41437117 on Build GRCh38
rs773490233 Chr.21: 41437015-41437015 on Build GRCh38
rs17000915 Chr.21: 41437118-41437118 on Build GRCh38
rs781581698 Chr.21: 41436989-41436989 on Build GRCh38
rs760453444 Chr.21: 41437019-41437019 on Build GRCh38
rs1003451642 Chr.21: 41437056-41437056 on Build GRCh38
rs764694357 Chr.21: 41437185-41437185 on Build GRCh38
rs775912670 Chr.21: 41436985-41436985 on Build GRCh38
rs372996528 Chr.21: 41437002-41437002 on Build GRCh38
rs775331020 Chr.21: 41437059-41437059 on Build GRCh38
rs764678342 Chr.21: 41437076-41437076 on Build GRCh38
rs377703833 Chr.21: 41437196-41437196 on Build GRCh38
rs868276196 Chr.21: 41437018-41437019 on Build GRCh38

View Full Product Details