Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41215926-41216181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHADL
Entrez Gene ID
150356
Gene Name
chondroadherin like
Gene Aliases
SLRR4B
UniGene
Hs.344488
Gene Chromosome Location
Chr.22: 41209101-41241773 on Build GRCh38
Transcripts
XM_005261370.3 XM_011529932.2 XM_011529934.2 NM_031488.4 XM_011530420.2 XM_011530421.2 XM_017028976.1 XM_017028977.1 XM_017028978.1 XM_017028979.1 XM_017028980.1

Gene Symbol
L3MBTL2
Entrez Gene ID
83746
Gene Name
L3MBTL2 polycomb repressive complex 1 subunit
Gene Aliases
H-l(3)mbt-l, L3MBT
UniGene
Hs.517641
Gene Chromosome Location
Chr.22: 41205192-41231271 on Build GRCh38
Transcripts
XM_005261370.3 XM_011529932.2 XM_011529934.2 NM_031488.4 XM_011530420.2 XM_011530421.2 XM_017028976.1 XM_017028977.1 XM_017028978.1 XM_017028979.1 XM_017028980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7279682 c.412delA p.A140fs*46 Chr.22: 41216154-41216154 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374600147 Chr.22: 41216108-41216108 on Build GRCh38
rs557489396 Chr.22: 41216077-41216077 on Build GRCh38
rs913506239 Chr.22: 41215972-41215972 on Build GRCh38
rs753730507 Chr.22: 41216126-41216126 on Build GRCh38
rs750903642 Chr.22: 41216099-41216099 on Build GRCh38
rs775015134 Chr.22: 41216049-41216049 on Build GRCh38
rs1028603046 Chr.22: 41215949-41215949 on Build GRCh38
rs369463912 Chr.22: 41216104-41216104 on Build GRCh38
rs763395713 Chr.22: 41216102-41216102 on Build GRCh38
rs982021667 Chr.22: 41216018-41216018 on Build GRCh38
rs967852555 Chr.22: 41216011-41216011 on Build GRCh38
rs538950201 Chr.22: 41216075-41216075 on Build GRCh38
rs775785110 Chr.22: 41215973-41215973 on Build GRCh38
rs567298031 Chr.22: 41215944-41215944 on Build GRCh38
rs546882298 Chr.22: 41216012-41216012 on Build GRCh38
rs777386965 Chr.22: 41216125-41216125 on Build GRCh38
rs990357167 Chr.22: 41215955-41215955 on Build GRCh38
rs946201035 Chr.22: 41215982-41215982 on Build GRCh38
rs374479045 Chr.22: 41215981-41215981 on Build GRCh38
rs142650987 Chr.22: 41216138-41216138 on Build GRCh38
rs974350564 Chr.22: 41215945-41215945 on Build GRCh38
rs928714805 Chr.22: 41216069-41216069 on Build GRCh38
rs571657118 Chr.22: 41216041-41216041 on Build GRCh38
rs528502700 Chr.22: 41215951-41215951 on Build GRCh38
rs757007045 Chr.22: 41216135-41216135 on Build GRCh38
rs778711090 Chr.22: 41216136-41216136 on Build GRCh38
rs767038856 Chr.22: 41216103-41216103 on Build GRCh38
rs1020317201 Chr.22: 41215974-41215975 on Build GRCh38
rs536318588 Chr.22: 41216150-41216150 on Build GRCh38

View Full Product Details