Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50359746-50359977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPSA
Entrez Gene ID
9476
Gene Name
napsin A aspartic peptidase
Gene Aliases
KAP, Kdap, NAP1, NAPA, SNAPA
UniGene
Hs.512843
Gene Chromosome Location
Chr.19: 50358472-50369331 on Build GRCh38
Transcripts
NM_004851.2 XM_011527540.1 XM_017027512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2754425 c.767C>T p.A256V Chr.19: 50359764-50359764 on Build GRCh38
3537185 c.747C>T p.F249F Chr.19: 50359784-50359784 on Build GRCh38
3757061 c.765T>C p.P255P Chr.19: 50359766-50359766 on Build GRCh38
5525286 c.763C>T p.P255S Chr.19: 50359768-50359768 on Build GRCh38
1395434 c.675T>C p.P225P Chr.19: 50359856-50359856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963476318 Chr.19: 50359836-50359836 on Build GRCh38
rs753918630 Chr.19: 50359868-50359868 on Build GRCh38
rs375232742 Chr.19: 50359848-50359848 on Build GRCh38
rs638294 Chr.19: 50359766-50359766 on Build GRCh38
rs773704054 Chr.19: 50359880-50359881 on Build GRCh38
rs528716175 Chr.19: 50359889-50359889 on Build GRCh38
rs748819177 Chr.19: 50359783-50359783 on Build GRCh38
rs761524390 Chr.19: 50359784-50359784 on Build GRCh38
rs201819380 Chr.19: 50359843-50359843 on Build GRCh38
rs778435688 Chr.19: 50359858-50359858 on Build GRCh38
rs143898038 Chr.19: 50359773-50359773 on Build GRCh38
rs756620162 Chr.19: 50359855-50359855 on Build GRCh38
rs634713 Chr.19: 50359928-50359928 on Build GRCh38
rs560915904 Chr.19: 50359887-50359887 on Build GRCh38
rs769339253 Chr.19: 50359913-50359913 on Build GRCh38
rs756132672 Chr.19: 50359800-50359800 on Build GRCh38
rs774523248 Chr.19: 50359820-50359821 on Build GRCh38
rs764878935 Chr.19: 50359785-50359785 on Build GRCh38
rs199991360 Chr.19: 50359875-50359875 on Build GRCh38
rs994659684 Chr.19: 50359767-50359767 on Build GRCh38
rs377299154 Chr.19: 50359811-50359811 on Build GRCh38
rs750124264 Chr.19: 50359840-50359840 on Build GRCh38
rs140973133 Chr.19: 50359821-50359821 on Build GRCh38
rs140131070 Chr.19: 50359772-50359772 on Build GRCh38
rs148925155 Chr.19: 50359812-50359812 on Build GRCh38
rs374700636 Chr.19: 50359809-50359809 on Build GRCh38
rs753293037 Chr.19: 50359797-50359797 on Build GRCh38
rs771942751 Chr.19: 50359816-50359816 on Build GRCh38
rs772647545 Chr.19: 50359861-50359861 on Build GRCh38
rs371222764 Chr.19: 50359803-50359803 on Build GRCh38
rs759794042 Chr.19: 50359841-50359841 on Build GRCh38
rs115708794 Chr.19: 50359817-50359817 on Build GRCh38
rs779968249 Chr.19: 50359882-50359882 on Build GRCh38
rs151042743 Chr.19: 50359820-50359820 on Build GRCh38
rs781517212 Chr.19: 50359796-50359796 on Build GRCh38
rs547179258 Chr.19: 50359897-50359897 on Build GRCh38
rs144934721 Chr.19: 50359878-50359878 on Build GRCh38
rs760724883 Chr.19: 50359818-50359818 on Build GRCh38
rs200649376 Chr.19: 50359844-50359844 on Build GRCh38
rs146890854 Chr.19: 50359793-50359793 on Build GRCh38
rs764825762 Chr.19: 50359837-50359837 on Build GRCh38
rs757443248 Chr.19: 50359869-50359869 on Build GRCh38
rs539333263 Chr.19: 50359825-50359830 on Build GRCh38
rs776576866 Chr.19: 50359896-50359896 on Build GRCh38
rs978664781 Chr.19: 50359908-50359908 on Build GRCh38
rs758116701 Chr.19: 50359881-50359881 on Build GRCh38
rs751816692 Chr.19: 50359787-50359787 on Build GRCh38

View Full Product Details