Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51719102-51719365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAS1
Entrez Gene ID
3036
Gene Name
hyaluronan synthase 1
Gene Aliases
HAS
UniGene
Hs.57697
Gene Chromosome Location
Chr.19: 51713112-51723992 on Build GRCh38
Transcripts
NM_001297436.1 NM_001523.3 XM_011526884.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
368715 c.635G>A p.W212* Chr.19: 51719273-51719273 on Build GRCh38
1000173 c.649G>A p.E217K Chr.19: 51719259-51719259 on Build GRCh38
1000172 c.679G>A p.G227R Chr.19: 51719229-51719229 on Build GRCh38
4435103 c.674C>T p.A225V Chr.19: 51719234-51719234 on Build GRCh38
3537876 c.582G>C p.L194L Chr.19: 51719326-51719326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932145096 Chr.19: 51719167-51719167 on Build GRCh38
rs17853475 Chr.19: 51719220-51719220 on Build GRCh38
rs200667452 Chr.19: 51719256-51719256 on Build GRCh38
rs909313293 Chr.19: 51719227-51719227 on Build GRCh38
rs750483953 Chr.19: 51719305-51719305 on Build GRCh38
rs781164597 Chr.19: 51719332-51719332 on Build GRCh38
rs769387541 Chr.19: 51719276-51719276 on Build GRCh38
rs761482836 Chr.19: 51719239-51719239 on Build GRCh38
rs779923824 Chr.19: 51719314-51719314 on Build GRCh38
rs769582481 Chr.19: 51719344-51719344 on Build GRCh38
rs768521219 Chr.19: 51719330-51719330 on Build GRCh38
rs945462052 Chr.19: 51719290-51719290 on Build GRCh38
rs747733820 Chr.19: 51719337-51719337 on Build GRCh38
rs756343125 Chr.19: 51719250-51719250 on Build GRCh38
rs144942434 Chr.19: 51719195-51719195 on Build GRCh38
rs746790040 Chr.19: 51719322-51719322 on Build GRCh38
rs568930180 Chr.19: 51719263-51719263 on Build GRCh38
rs753014604 Chr.19: 51719248-51719248 on Build GRCh38
rs775182179 Chr.19: 51719211-51719211 on Build GRCh38
rs370795750 Chr.19: 51719286-51719286 on Build GRCh38
rs749475490 Chr.19: 51719191-51719191 on Build GRCh38
rs374038790 Chr.19: 51719277-51719277 on Build GRCh38
rs550695209 Chr.19: 51719229-51719229 on Build GRCh38
rs767853866 Chr.19: 51719247-51719247 on Build GRCh38
rs770847948 Chr.19: 51719182-51719182 on Build GRCh38
rs11558146 Chr.19: 51719237-51719237 on Build GRCh38
rs11084112 Chr.19: 51719299-51719299 on Build GRCh38
rs1054195799 Chr.19: 51719255-51719255 on Build GRCh38
rs753097128 Chr.19: 51719288-51719288 on Build GRCh38
rs745632765 Chr.19: 51719186-51719186 on Build GRCh38
rs148796716 Chr.19: 51719270-51719270 on Build GRCh38
rs1032740488 Chr.19: 51719329-51719329 on Build GRCh38
rs778990103 Chr.19: 51719257-51719257 on Build GRCh38
rs370617787 Chr.19: 51719269-51719269 on Build GRCh38
rs755469539 Chr.19: 51719246-51719246 on Build GRCh38
rs949986992 Chr.19: 51719262-51719262 on Build GRCh38
rs774316728 Chr.19: 51719185-51719185 on Build GRCh38
rs764282361 Chr.19: 51719296-51719296 on Build GRCh38
rs778987027 Chr.19: 51719302-51719302 on Build GRCh38
rs774926345 Chr.19: 51719212-51719212 on Build GRCh38
rs761579314 Chr.19: 51719273-51719273 on Build GRCh38
rs771909540 Chr.19: 51719261-51719261 on Build GRCh38
rs745780732 Chr.19: 51719260-51719260 on Build GRCh38
rs375831712 Chr.19: 51719280-51719280 on Build GRCh38
rs778031457 Chr.19: 51719251-51719251 on Build GRCh38
rs757504132 Chr.19: 51719301-51719301 on Build GRCh38
rs555126388 Chr.19: 51719243-51719243 on Build GRCh38
rs773113588 Chr.19: 51719195-51719196 on Build GRCh38
rs912723656 Chr.19: 51719218-51719218 on Build GRCh38
rs569217641 Chr.19: 51719169-51719169 on Build GRCh38
rs761064565 Chr.19: 51719292-51719292 on Build GRCh38
rs758345819 Chr.19: 51719308-51719308 on Build GRCh38
rs144360076 Chr.19: 51719252-51719252 on Build GRCh38

View Full Product Details