Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55822516-55822785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC14
Entrez Gene ID
85406
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Gene Aliases
DNAJ, DRIP78, HDJ3, LIP6
UniGene
Hs.709320
Gene Chromosome Location
Chr.12: 55820960-55829636 on Build GRCh38
Transcripts
NM_032364.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5572752 c.1757A>G p.Y586C Chr.12: 55822610-55822610 on Build GRCh38
5015122 c.1675T>C p.C559R Chr.12: 55822692-55822692 on Build GRCh38
6332617 c.1783T>C p.Y595H Chr.12: 55822584-55822584 on Build GRCh38
5841467 c.1741G>A p.G581S Chr.12: 55822626-55822626 on Build GRCh38
6647256 c.1652G>T p.R551L Chr.12: 55822715-55822715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781539452 Chr.12: 55822685-55822685 on Build GRCh38
rs758907773 Chr.12: 55822603-55822603 on Build GRCh38
rs753341713 Chr.12: 55822574-55822574 on Build GRCh38
rs534529653 Chr.12: 55822716-55822716 on Build GRCh38
rs745615260 Chr.12: 55822619-55822619 on Build GRCh38
rs769499414 Chr.12: 55822677-55822677 on Build GRCh38
rs1015970656 Chr.12: 55822631-55822631 on Build GRCh38
rs570693847 Chr.12: 55822687-55822687 on Build GRCh38
rs775617259 Chr.12: 55822713-55822713 on Build GRCh38
rs996310464 Chr.12: 55822693-55822693 on Build GRCh38
rs377323880 Chr.12: 55822562-55822562 on Build GRCh38
rs999769176 Chr.12: 55822747-55822747 on Build GRCh38
rs747575227 Chr.12: 55822660-55822660 on Build GRCh38
rs767419303 Chr.12: 55822750-55822750 on Build GRCh38
rs774105541 Chr.12: 55822738-55822738 on Build GRCh38
rs778101506 Chr.12: 55822629-55822629 on Build GRCh38
rs370435412 Chr.12: 55822578-55822578 on Build GRCh38
rs930586916 Chr.12: 55822604-55822604 on Build GRCh38
rs928342684 Chr.12: 55822756-55822756 on Build GRCh38
rs1047421032 Chr.12: 55822630-55822630 on Build GRCh38
rs553087983 Chr.12: 55822754-55822754 on Build GRCh38
rs554043997 Chr.12: 55822563-55822564 on Build GRCh38
rs758545015 Chr.12: 55822639-55822640 on Build GRCh38
rs768808249 Chr.12: 55822730-55822730 on Build GRCh38
rs771228489 Chr.12: 55822674-55822674 on Build GRCh38
rs969697059 Chr.12: 55822635-55822635 on Build GRCh38
rs944453956 Chr.12: 55822676-55822676 on Build GRCh38
rs755899802 Chr.12: 55822544-55822544 on Build GRCh38
rs770059143 Chr.12: 55822696-55822696 on Build GRCh38
rs200567712 Chr.12: 55822740-55822740 on Build GRCh38

View Full Product Details