Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26198690-26198961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2AD
Entrez Gene ID
3013
Gene Name
histone cluster 1, H2ad
Gene Aliases
H2A.3, H2A/g, H2AFG
UniGene
Hs.626666
Gene Chromosome Location
Chr.6: 26198784-26199293 on Build GRCh38
Transcripts
NM_021065.3 NM_003530.4

Gene Symbol
HIST1H3D
Entrez Gene ID
8351
Gene Name
histone cluster 1, H3d
Gene Aliases
H3/b, H3FB
UniGene
Hs.532144
Gene Chromosome Location
Chr.6: 26196784-26199293 on Build GRCh38
Transcripts
NM_021065.3 NM_003530.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
451021 c.364G>C p.E122Q Chr.6: 26198880-26198880 on Build GRCh38
3860925 c.360G>C p.K120N Chr.6: 26198884-26198884 on Build GRCh38
351363 c.383A>G p.K128R Chr.6: 26198861-26198861 on Build GRCh38
3745094 c.365A>T p.E122V Chr.6: 26198879-26198879 on Build GRCh38
7109412 c.340G>C p.A114P Chr.6: 26198904-26198904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112920691 Chr.6: 26198935-26198935 on Build GRCh38
rs752532053 Chr.6: 26198874-26198874 on Build GRCh38
rs112070326 Chr.6: 26198821-26198821 on Build GRCh38
rs749033364 Chr.6: 26198901-26198901 on Build GRCh38
rs558541909 Chr.6: 26198831-26198831 on Build GRCh38
rs77463023 Chr.6: 26198752-26198752 on Build GRCh38
rs765769347 Chr.6: 26198856-26198856 on Build GRCh38
rs372679752 Chr.6: 26198813-26198813 on Build GRCh38
rs747684713 Chr.6: 26198817-26198817 on Build GRCh38
rs371620303 Chr.6: 26198764-26198764 on Build GRCh38
rs945070951 Chr.6: 26198771-26198771 on Build GRCh38
rs1021992523 Chr.6: 26198784-26198784 on Build GRCh38
rs755558538 Chr.6: 26198802-26198802 on Build GRCh38
rs145059830 Chr.6: 26198893-26198893 on Build GRCh38
rs1003050974 Chr.6: 26198736-26198736 on Build GRCh38
rs770198458 Chr.6: 26198912-26198912 on Build GRCh38
rs749812440 Chr.6: 26198914-26198914 on Build GRCh38
rs781111556 Chr.6: 26198895-26198895 on Build GRCh38
rs772797188 Chr.6: 26198853-26198853 on Build GRCh38
rs760519610 Chr.6: 26198859-26198864 on Build GRCh38
rs561037127 Chr.6: 26198865-26198865 on Build GRCh38
rs536423288 Chr.6: 26198761-26198761 on Build GRCh38
rs34550794 Chr.6: 26198786-26198786 on Build GRCh38
rs530347788 Chr.6: 26198878-26198878 on Build GRCh38
rs914864615 Chr.6: 26198770-26198770 on Build GRCh38
rs777723358 Chr.6: 26198884-26198884 on Build GRCh38
rs141019559 Chr.6: 26198727-26198727 on Build GRCh38
rs768934419 Chr.6: 26198880-26198881 on Build GRCh38
rs142041097 Chr.6: 26198938-26198938 on Build GRCh38
rs1015789599 Chr.6: 26198866-26198866 on Build GRCh38
rs749932231 Chr.6: 26198800-26198800 on Build GRCh38
rs887806856 Chr.6: 26198887-26198887 on Build GRCh38
rs780158998 Chr.6: 26198834-26198835 on Build GRCh38
rs1005162636 Chr.6: 26198723-26198723 on Build GRCh38
rs753426007 Chr.6: 26198859-26198859 on Build GRCh38
rs921757485 Chr.6: 26198819-26198819 on Build GRCh38
rs932119618 Chr.6: 26198832-26198832 on Build GRCh38
rs1015165682 Chr.6: 26198726-26198726 on Build GRCh38
rs768963286 Chr.6: 26198915-26198915 on Build GRCh38
rs774731285 Chr.6: 26198919-26198919 on Build GRCh38
rs759367395 Chr.6: 26198864-26198864 on Build GRCh38
rs538661204 Chr.6: 26198830-26198830 on Build GRCh38
rs573140623 Chr.6: 26198808-26198808 on Build GRCh38
rs749358448 Chr.6: 26198876-26198877 on Build GRCh38
rs780472934 Chr.6: 26198913-26198913 on Build GRCh38
rs765273519 Chr.6: 26198870-26198870 on Build GRCh38
rs770822366 Chr.6: 26198934-26198934 on Build GRCh38
rs756050081 Chr.6: 26198825-26198826 on Build GRCh38
rs376397894 Chr.6: 26198835-26198835 on Build GRCh38
rs977706074 Chr.6: 26198787-26198787 on Build GRCh38
rs1042291244 Chr.6: 26198774-26198774 on Build GRCh38
rs760217565 Chr.6: 26198932-26198932 on Build GRCh38
rs879066162 Chr.6: 26198779-26198779 on Build GRCh38
rs111373540 Chr.6: 26198794-26198794 on Build GRCh38
rs770663132 Chr.6: 26198933-26198933 on Build GRCh38
rs760402571 Chr.6: 26198854-26198854 on Build GRCh38
rs548651970 Chr.6: 26198745-26198745 on Build GRCh38
rs769014121 Chr.6: 26198847-26198847 on Build GRCh38
rs754117574 Chr.6: 26198729-26198729 on Build GRCh38
rs750883078 Chr.6: 26198773-26198773 on Build GRCh38
rs780806744 Chr.6: 26198814-26198814 on Build GRCh38
rs34598107 Chr.6: 26198737-26198738 on Build GRCh38
rs751780330 Chr.6: 26198888-26198888 on Build GRCh38
rs76827793 Chr.6: 26198836-26198836 on Build GRCh38
rs770383182 Chr.6: 26198842-26198842 on Build GRCh38
rs926361318 Chr.6: 26198733-26198733 on Build GRCh38
rs544017356 Chr.6: 26198840-26198840 on Build GRCh38
rs575644929 Chr.6: 26198726-26198729 on Build GRCh38
rs775821141 Chr.6: 26198845-26198845 on Build GRCh38
rs796781483 Chr.6: 26198880-26198881 on Build GRCh38

View Full Product Details