Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49459070-49459317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6
Entrez Gene ID
2074
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Gene Aliases
ARMD5, CKN2, COFS, COFS1, CSB, POF11, RAD26, UVSS1
UniGene
Hs.49063
Gene Chromosome Location
Chr.10: 49454480-49539538 on Build GRCh38
Transcripts
NM_000124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665615 c.4150delC p.L1384fs*10 Chr.10: 49459147-49459147 on Build GRCh38
127906 c.4122A>G p.A1374A Chr.10: 49459175-49459175 on Build GRCh38
1348065 c.4203G>A p.L1401L Chr.10: 49459094-49459094 on Build GRCh38
216221 c.4211G>A p.R1404H Chr.10: 49459086-49459086 on Build GRCh38
538877 c.4143C>T p.S1381S Chr.10: 49459154-49459154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781538914 Chr.10: 49459168-49459168 on Build GRCh38
rs776383043 Chr.10: 49459222-49459222 on Build GRCh38
rs4253227 Chr.10: 49459183-49459183 on Build GRCh38
rs985847612 Chr.10: 49459253-49459253 on Build GRCh38
rs4253228 Chr.10: 49459153-49459153 on Build GRCh38
rs751666822 Chr.10: 49459157-49459157 on Build GRCh38
rs578171795 Chr.10: 49459257-49459257 on Build GRCh38
rs773819856 Chr.10: 49459117-49459117 on Build GRCh38
rs965697729 Chr.10: 49459285-49459285 on Build GRCh38
rs55838288 Chr.10: 49459220-49459220 on Build GRCh38
rs766614166 Chr.10: 49459242-49459244 on Build GRCh38
rs34917815 Chr.10: 49459232-49459232 on Build GRCh38
rs761551318 Chr.10: 49459170-49459172 on Build GRCh38
rs766980240 Chr.10: 49459235-49459235 on Build GRCh38
rs961974385 Chr.10: 49459108-49459108 on Build GRCh38
rs750916757 Chr.10: 49459147-49459147 on Build GRCh38
rs771532766 Chr.10: 49459113-49459113 on Build GRCh38
rs762286899 Chr.10: 49459143-49459143 on Build GRCh38
rs765804876 Chr.10: 49459145-49459145 on Build GRCh38
rs755154783 Chr.10: 49459163-49459163 on Build GRCh38
rs764956689 Chr.10: 49459274-49459274 on Build GRCh38
rs370285377 Chr.10: 49459230-49459230 on Build GRCh38
rs1043213652 Chr.10: 49459151-49459151 on Build GRCh38
rs753553933 Chr.10: 49459170-49459170 on Build GRCh38
rs747021788 Chr.10: 49459249-49459249 on Build GRCh38
rs746586048 Chr.10: 49459203-49459203 on Build GRCh38
rs375921453 Chr.10: 49459154-49459154 on Build GRCh38
rs370123825 Chr.10: 49459252-49459252 on Build GRCh38
rs775089627 Chr.10: 49459123-49459123 on Build GRCh38
rs892019345 Chr.10: 49459112-49459112 on Build GRCh38
rs574272317 Chr.10: 49459231-49459231 on Build GRCh38
rs745352643 Chr.10: 49459111-49459111 on Build GRCh38
rs373553169 Chr.10: 49459265-49459265 on Build GRCh38
rs568838614 Chr.10: 49459193-49459193 on Build GRCh38
rs150277901 Chr.10: 49459202-49459202 on Build GRCh38
rs114694744 Chr.10: 49459139-49459139 on Build GRCh38
rs752925338 Chr.10: 49459261-49459261 on Build GRCh38
rs369444705 Chr.10: 49459137-49459137 on Build GRCh38
rs186482480 Chr.10: 49459239-49459239 on Build GRCh38
rs745758957 Chr.10: 49459201-49459201 on Build GRCh38
rs1055848255 Chr.10: 49459160-49459160 on Build GRCh38
rs754224226 Chr.10: 49459282-49459283 on Build GRCh38

View Full Product Details