Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47341610-47341845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5AR2
Entrez Gene ID
27202
Gene Name
complement component 5a receptor 2
Gene Aliases
C5L2, GPF77, GPR77
UniGene
Hs.534412
Gene Chromosome Location
Chr.19: 47331614-47342080 on Build GRCh38
Transcripts
NM_001271749.1 NM_001271750.1 NM_018485.2 XM_011526736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712534 c.835C>T p.L279F Chr.19: 47341634-47341634 on Build GRCh38
3713140 c.815C>A p.A272D Chr.19: 47341614-47341614 on Build GRCh38
4856000 c.892C>G p.Q298E Chr.19: 47341691-47341691 on Build GRCh38
6616237 c.840C>T p.A280A Chr.19: 47341639-47341639 on Build GRCh38
5591150 c.841C>T p.L281F Chr.19: 47341640-47341640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759447410 Chr.19: 47341676-47341676 on Build GRCh38
rs773403648 Chr.19: 47341774-47341774 on Build GRCh38
rs780336750 Chr.19: 47341708-47341708 on Build GRCh38
rs200496715 Chr.19: 47341778-47341778 on Build GRCh38
rs147408302 Chr.19: 47341697-47341697 on Build GRCh38
rs373468449 Chr.19: 47341809-47341809 on Build GRCh38
rs776640499 Chr.19: 47341768-47341768 on Build GRCh38
rs150599989 Chr.19: 47341767-47341767 on Build GRCh38
rs112290646 Chr.19: 47341785-47341785 on Build GRCh38
rs769576418 Chr.19: 47341645-47341645 on Build GRCh38
rs148463474 Chr.19: 47341700-47341700 on Build GRCh38
rs200192224 Chr.19: 47341643-47341643 on Build GRCh38
rs768162059 Chr.19: 47341788-47341788 on Build GRCh38
rs758762136 Chr.19: 47341701-47341701 on Build GRCh38
rs764519595 Chr.19: 47341769-47341769 on Build GRCh38
rs1045465534 Chr.19: 47341629-47341629 on Build GRCh38
rs369783726 Chr.19: 47341797-47341797 on Build GRCh38
rs780687004 Chr.19: 47341740-47341740 on Build GRCh38
rs774323355 Chr.19: 47341667-47341667 on Build GRCh38
rs764228335 Chr.19: 47341689-47341689 on Build GRCh38
rs373635111 Chr.19: 47341729-47341729 on Build GRCh38
rs777481847 Chr.19: 47341798-47341798 on Build GRCh38
rs758850692 Chr.19: 47341628-47341628 on Build GRCh38
rs752215226 Chr.19: 47341793-47341793 on Build GRCh38
rs971673684 Chr.19: 47341794-47341794 on Build GRCh38
rs747676401 Chr.19: 47341741-47341741 on Build GRCh38
rs199778653 Chr.19: 47341662-47341662 on Build GRCh38
rs1038541607 Chr.19: 47341672-47341672 on Build GRCh38
rs761910522 Chr.19: 47341771-47341771 on Build GRCh38
rs773084653 Chr.19: 47341646-47341646 on Build GRCh38
rs141780251 Chr.19: 47341753-47341753 on Build GRCh38
rs774081017 Chr.19: 47341765-47341765 on Build GRCh38
rs766765736 Chr.19: 47341775-47341775 on Build GRCh38
rs202152698 Chr.19: 47341632-47341632 on Build GRCh38
rs34085015 Chr.19: 47341642-47341642 on Build GRCh38
rs777535147 Chr.19: 47341744-47341744 on Build GRCh38
rs750743266 Chr.19: 47341699-47341699 on Build GRCh38
rs139142067 Chr.19: 47341657-47341657 on Build GRCh38
rs117238101 Chr.19: 47341698-47341698 on Build GRCh38
rs376929195 Chr.19: 47341820-47341820 on Build GRCh38
rs780645967 Chr.19: 47341640-47341640 on Build GRCh38
rs200456413 Chr.19: 47341664-47341664 on Build GRCh38
rs755504740 Chr.19: 47341735-47341735 on Build GRCh38
rs143696467 Chr.19: 47341743-47341743 on Build GRCh38
rs529217199 Chr.19: 47341754-47341754 on Build GRCh38
rs961239509 Chr.19: 47341707-47341707 on Build GRCh38
rs142782206 Chr.19: 47341694-47341694 on Build GRCh38

View Full Product Details