Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31580626-31580897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTB
Entrez Gene ID
4050
Gene Name
lymphotoxin beta
Gene Aliases
TNFC, TNFSF3, TNLG1C, p33
UniGene
Hs.376208
Gene Chromosome Location
Chr.6: 31580558-31582425 on Build GRCh38
Transcripts
NM_002341.1 NM_009588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6173656 c.586G>T p.G196W Chr.6: 31580858-31580858 on Build GRCh38
4554276 c.616G>A p.G206R Chr.6: 31580828-31580828 on Build GRCh38
3253339 c.679G>A p.D227N Chr.6: 31580765-31580765 on Build GRCh38
290670 c.624C>T p.G208G Chr.6: 31580820-31580820 on Build GRCh38
1443286 c.719C>T p.A240V Chr.6: 31580725-31580725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747146913 Chr.6: 31580724-31580724 on Build GRCh38
rs556812665 Chr.6: 31580847-31580847 on Build GRCh38
rs4248165 Chr.6: 31580751-31580751 on Build GRCh38
rs768807517 Chr.6: 31580678-31580679 on Build GRCh38
rs771169298 Chr.6: 31580727-31580727 on Build GRCh38
rs145227210 Chr.6: 31580766-31580766 on Build GRCh38
rs1049718572 Chr.6: 31580837-31580837 on Build GRCh38
rs748549211 Chr.6: 31580868-31580868 on Build GRCh38
rs746871696 Chr.6: 31580783-31580783 on Build GRCh38
rs779671727 Chr.6: 31580665-31580665 on Build GRCh38
rs149139835 Chr.6: 31580809-31580809 on Build GRCh38
rs759586959 Chr.6: 31580765-31580765 on Build GRCh38
rs748760498 Chr.6: 31580686-31580686 on Build GRCh38
rs879158785 Chr.6: 31580764-31580764 on Build GRCh38
rs754586202 Chr.6: 31580673-31580673 on Build GRCh38
rs966815487 Chr.6: 31580660-31580660 on Build GRCh38
rs750545997 Chr.6: 31580876-31580876 on Build GRCh38
rs377440823 Chr.6: 31580697-31580697 on Build GRCh38
rs773644231 Chr.6: 31580713-31580713 on Build GRCh38
rs756014417 Chr.6: 31580816-31580816 on Build GRCh38
rs772262931 Chr.6: 31580687-31580687 on Build GRCh38
rs534349570 Chr.6: 31580661-31580661 on Build GRCh38
rs754534730 Chr.6: 31580850-31580850 on Build GRCh38
rs777955797 Chr.6: 31580873-31580873 on Build GRCh38
rs575140707 Chr.6: 31580878-31580878 on Build GRCh38
rs748778397 Chr.6: 31580667-31580667 on Build GRCh38
rs930822365 Chr.6: 31580833-31580833 on Build GRCh38
rs761813329 Chr.6: 31580774-31580774 on Build GRCh38
rs778494063 Chr.6: 31580867-31580867 on Build GRCh38
rs1014107548 Chr.6: 31580777-31580777 on Build GRCh38
rs3093556 Chr.6: 31580674-31580674 on Build GRCh38
rs766109943 Chr.6: 31580822-31580822 on Build GRCh38
rs776875834 Chr.6: 31580748-31580748 on Build GRCh38

View Full Product Details