Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156705272-156705529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRABP2
Entrez Gene ID
1382
Gene Name
cellular retinoic acid binding protein 2
Gene Aliases
CRABP-II, RBP6
UniGene
Hs.405662
Gene Chromosome Location
Chr.1: 156699606-156712900 on Build GRCh38
Transcripts
NM_001199723.1 NM_001878.3 XM_017000343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4023383 c.58C>T p.L20F Chr.1: 156705389-156705389 on Build GRCh38
1984142 c.25A>G p.K9E Chr.1: 156705422-156705422 on Build GRCh38
4674565 c.27A>G p.K9K Chr.1: 156705420-156705420 on Build GRCh38
6459410 c.5C>T p.P2L Chr.1: 156705442-156705442 on Build GRCh38
4622161 c.34C>T p.R12* Chr.1: 156705413-156705413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146310086 Chr.1: 156705387-156705387 on Build GRCh38
rs780773653 Chr.1: 156705453-156705453 on Build GRCh38
rs546957711 Chr.1: 156705379-156705379 on Build GRCh38
rs917116388 Chr.1: 156705451-156705451 on Build GRCh38
rs770917529 Chr.1: 156705358-156705358 on Build GRCh38
rs779424358 Chr.1: 156705330-156705330 on Build GRCh38
rs376300234 Chr.1: 156705326-156705326 on Build GRCh38
rs979923752 Chr.1: 156705503-156705503 on Build GRCh38
rs765195704 Chr.1: 156705408-156705408 on Build GRCh38
rs764322383 Chr.1: 156705389-156705389 on Build GRCh38
rs11590924 Chr.1: 156705353-156705353 on Build GRCh38
rs758958941 Chr.1: 156705418-156705418 on Build GRCh38
rs775500633 Chr.1: 156705487-156705487 on Build GRCh38
rs754025759 Chr.1: 156705398-156705398 on Build GRCh38
rs201336739 Chr.1: 156705461-156705461 on Build GRCh38
rs750577120 Chr.1: 156705413-156705413 on Build GRCh38
rs374879624 Chr.1: 156705367-156705367 on Build GRCh38
rs745393005 Chr.1: 156705336-156705337 on Build GRCh38
rs769221456 Chr.1: 156705350-156705350 on Build GRCh38
rs748252487 Chr.1: 156705319-156705319 on Build GRCh38
rs978559038 Chr.1: 156705392-156705392 on Build GRCh38
rs988754810 Chr.1: 156705388-156705388 on Build GRCh38
rs755310208 Chr.1: 156705472-156705472 on Build GRCh38
rs200159911 Chr.1: 156705341-156705341 on Build GRCh38
rs780540644 Chr.1: 156705338-156705338 on Build GRCh38
rs780858726 Chr.1: 156705495-156705495 on Build GRCh38
rs968479613 Chr.1: 156705390-156705390 on Build GRCh38
rs750995481 Chr.1: 156705337-156705337 on Build GRCh38
rs762856531 Chr.1: 156705357-156705357 on Build GRCh38
rs867628673 Chr.1: 156705396-156705396 on Build GRCh38
rs536726294 Chr.1: 156705366-156705366 on Build GRCh38
rs367677751 Chr.1: 156705342-156705342 on Build GRCh38
rs771650678 Chr.1: 156705337-156705337 on Build GRCh38
rs774132730 Chr.1: 156705377-156705377 on Build GRCh38
rs747435489 Chr.1: 156705339-156705339 on Build GRCh38
rs761911054 Chr.1: 156705399-156705399 on Build GRCh38

View Full Product Details