Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1816690-1816963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LETM1
Entrez Gene ID
3954
Gene Name
leucine zipper and EF-hand containing transmembrane protein 1
Gene Aliases
-
UniGene
Hs.120165
Gene Chromosome Location
Chr.4: 1811479-1856247 on Build GRCh38
Transcripts
NM_012318.2 XM_006713884.1 XM_017008187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866104 c.1837C>T p.Q613* Chr.4: 1816821-1816821 on Build GRCh38
6299320 c.1817A>T p.K606M Chr.4: 1816841-1816841 on Build GRCh38
6700584 c.1917C>T p.N639N Chr.4: 1816741-1816741 on Build GRCh38
2157263 c.1744-1G>A p.? Chr.4: 1816915-1816915 on Build GRCh38
1428758 c.1744G>A p.D582N Chr.4: 1816914-1816914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373654818 Chr.4: 1816809-1816809 on Build GRCh38
rs201689850 Chr.4: 1816825-1816825 on Build GRCh38
rs894669104 Chr.4: 1816860-1816860 on Build GRCh38
rs537070207 Chr.4: 1816730-1816730 on Build GRCh38
rs112963620 Chr.4: 1816894-1816894 on Build GRCh38
rs779574558 Chr.4: 1816771-1816771 on Build GRCh38
rs779486422 Chr.4: 1816861-1816861 on Build GRCh38
rs756578993 Chr.4: 1816863-1816863 on Build GRCh38
rs758054072 Chr.4: 1816846-1816846 on Build GRCh38
rs532051187 Chr.4: 1816836-1816836 on Build GRCh38
rs767723941 Chr.4: 1816732-1816732 on Build GRCh38
rs773573307 Chr.4: 1816717-1816717 on Build GRCh38
rs764194986 Chr.4: 1816826-1816826 on Build GRCh38
rs181682155 Chr.4: 1816729-1816729 on Build GRCh38
rs753632622 Chr.4: 1816741-1816741 on Build GRCh38
rs748591725 Chr.4: 1816785-1816785 on Build GRCh38
rs372297853 Chr.4: 1816714-1816714 on Build GRCh38
rs535970886 Chr.4: 1816796-1816796 on Build GRCh38
rs201582550 Chr.4: 1816707-1816707 on Build GRCh38
rs763264804 Chr.4: 1816801-1816801 on Build GRCh38
rs908472279 Chr.4: 1816733-1816733 on Build GRCh38
rs115233072 Chr.4: 1816898-1816898 on Build GRCh38
rs750006587 Chr.4: 1816843-1816843 on Build GRCh38
rs769120520 Chr.4: 1816709-1816709 on Build GRCh38
rs763180183 Chr.4: 1816726-1816726 on Build GRCh38
rs530212467 Chr.4: 1816918-1816918 on Build GRCh38
rs753637801 Chr.4: 1816830-1816830 on Build GRCh38
rs202160762 Chr.4: 1816744-1816744 on Build GRCh38
rs770108174 Chr.4: 1816716-1816716 on Build GRCh38
rs376355543 Chr.4: 1816736-1816736 on Build GRCh38
rs771157898 Chr.4: 1816803-1816803 on Build GRCh38
rs531230067 Chr.4: 1816745-1816745 on Build GRCh38
rs762018003 Chr.4: 1816908-1816908 on Build GRCh38
rs746028144 Chr.4: 1816897-1816897 on Build GRCh38
rs767637991 Chr.4: 1816810-1816810 on Build GRCh38
rs777172217 Chr.4: 1816713-1816713 on Build GRCh38
rs771537649 Chr.4: 1816786-1816786 on Build GRCh38
rs556747102 Chr.4: 1816735-1816735 on Build GRCh38
rs776385710 Chr.4: 1816927-1816927 on Build GRCh38
rs762334176 Chr.4: 1816715-1816715 on Build GRCh38
rs771522729 Chr.4: 1816748-1816748 on Build GRCh38
rs373257565 Chr.4: 1816708-1816708 on Build GRCh38
rs879062041 Chr.4: 1816727-1816727 on Build GRCh38
rs1007896822 Chr.4: 1816858-1816858 on Build GRCh38
rs9328763 Chr.4: 1816867-1816867 on Build GRCh38
rs761843561 Chr.4: 1816930-1816930 on Build GRCh38
rs777651583 Chr.4: 1816740-1816740 on Build GRCh38
rs115591912 Chr.4: 1816841-1816841 on Build GRCh38
rs371042512 Chr.4: 1816747-1816747 on Build GRCh38
rs778189000 Chr.4: 1816864-1816864 on Build GRCh38
rs774383747 Chr.4: 1816808-1816808 on Build GRCh38
rs760879319 Chr.4: 1816912-1816912 on Build GRCh38
rs996630944 Chr.4: 1816777-1816777 on Build GRCh38

View Full Product Details