Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 449997-450234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_011534748.2 XM_011534749.1 XM_017013313.1 XM_017013314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7351760 c.475+2T>C p.? Chr.8: 450085-450085 on Build GRCh38
3900133 c.426C>T p.G142G Chr.8: 450034-450034 on Build GRCh38
6377495 c.415T>C p.S139P Chr.8: 450023-450023 on Build GRCh38
6662143 c.467T>C p.V156A Chr.8: 450075-450075 on Build GRCh38
1206820 c.427G>A p.V143M Chr.8: 450035-450035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545238483 Chr.8: 450141-450141 on Build GRCh38
rs775999717 Chr.8: 450050-450050 on Build GRCh38
rs764111651 Chr.8: 450056-450056 on Build GRCh38
rs1038679282 Chr.8: 450144-450144 on Build GRCh38
rs397841700 Chr.8: 450203-450205 on Build GRCh38
rs924431208 Chr.8: 450109-450109 on Build GRCh38
rs1024971631 Chr.8: 450206-450206 on Build GRCh38
rs749780986 Chr.8: 450131-450131 on Build GRCh38
rs760671974 Chr.8: 450189-450189 on Build GRCh38
rs770069942 Chr.8: 450042-450042 on Build GRCh38
rs900485039 Chr.8: 450156-450156 on Build GRCh38
rs143947386 Chr.8: 450186-450186 on Build GRCh38
rs146187216 Chr.8: 450124-450124 on Build GRCh38
rs144100418 Chr.8: 450057-450057 on Build GRCh38
rs562354345 Chr.8: 450074-450074 on Build GRCh38
rs111862296 Chr.8: 450202-450203 on Build GRCh38
rs1008028929 Chr.8: 450192-450192 on Build GRCh38
rs959634440 Chr.8: 450193-450193 on Build GRCh38
rs34715204 Chr.8: 450139-450140 on Build GRCh38
rs182390506 Chr.8: 450161-450161 on Build GRCh38
rs189931167 Chr.8: 450123-450123 on Build GRCh38
rs754983811 Chr.8: 450099-450099 on Build GRCh38
rs949477474 Chr.8: 450071-450071 on Build GRCh38
rs991250154 Chr.8: 450197-450197 on Build GRCh38
rs3841577 Chr.8: 450200-450202 on Build GRCh38
rs753765477 Chr.8: 450096-450096 on Build GRCh38
rs750173761 Chr.8: 450075-450075 on Build GRCh38
rs997215305 Chr.8: 450150-450150 on Build GRCh38
rs777538570 Chr.8: 450112-450112 on Build GRCh38
rs918182966 Chr.8: 450073-450073 on Build GRCh38
rs533332261 Chr.8: 450136-450136 on Build GRCh38
rs200998638 Chr.8: 450199-450201 on Build GRCh38
rs187097378 Chr.8: 450169-450169 on Build GRCh38
rs897029175 Chr.8: 450201-450201 on Build GRCh38
rs763402699 Chr.8: 450052-450052 on Build GRCh38
rs761994781 Chr.8: 450064-450064 on Build GRCh38
rs934447475 Chr.8: 450122-450122 on Build GRCh38
rs149484405 Chr.8: 450149-450149 on Build GRCh38
rs777103868 Chr.8: 450033-450033 on Build GRCh38
rs760458112 Chr.8: 450087-450087 on Build GRCh38
rs778543895 Chr.8: 450100-450100 on Build GRCh38
rs775365943 Chr.8: 450090-450091 on Build GRCh38
rs2290073 Chr.8: 450168-450168 on Build GRCh38
rs370178351 Chr.8: 450116-450116 on Build GRCh38
rs185946180 Chr.8: 450034-450034 on Build GRCh38
rs918022123 Chr.8: 450058-450058 on Build GRCh38
rs137862229 Chr.8: 450128-450128 on Build GRCh38
rs758118143 Chr.8: 450106-450106 on Build GRCh38
rs752294810 Chr.8: 450104-450104 on Build GRCh38
rs766089971 Chr.8: 450095-450095 on Build GRCh38

View Full Product Details