Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241500532-241500769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FH
Entrez Gene ID
2271
Gene Name
fumarate hydratase
Gene Aliases
FMRD, HLRCC, LRCC, MCL, MCUL1
UniGene
Hs.592490
Gene Chromosome Location
Chr.1: 241497557-241519785 on Build GRCh38
Transcripts
NM_000143.3 XM_011544132.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731414 c.1273G>T p.D425Y Chr.1: 241500554-241500554 on Build GRCh38
6424532 c.1292C>T p.T431I Chr.1: 241500535-241500535 on Build GRCh38
350010 c.1280C>T p.S427L Chr.1: 241500547-241500547 on Build GRCh38
679717 c.1246G>C p.V416L Chr.1: 241500581-241500581 on Build GRCh38
5017765 c.1273delG p.D425fs*24 Chr.1: 241500554-241500554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200490916 Chr.1: 241500624-241500624 on Build GRCh38
rs969575206 Chr.1: 241500707-241500707 on Build GRCh38
rs190374342 Chr.1: 241500636-241500636 on Build GRCh38
rs980915467 Chr.1: 241500728-241500728 on Build GRCh38
rs1030047090 Chr.1: 241500670-241500670 on Build GRCh38
rs201765715 Chr.1: 241500635-241500635 on Build GRCh38
rs201422946 Chr.1: 241500639-241500639 on Build GRCh38
rs892493783 Chr.1: 241500643-241500643 on Build GRCh38
rs748406682 Chr.1: 241500604-241500605 on Build GRCh38
rs376260223 Chr.1: 241500597-241500597 on Build GRCh38
rs779985493 Chr.1: 241500598-241500599 on Build GRCh38
rs777822108 Chr.1: 241500617-241500617 on Build GRCh38
rs543506404 Chr.1: 241500724-241500724 on Build GRCh38
rs765639208 Chr.1: 241500593-241500594 on Build GRCh38
rs775301109 Chr.1: 241500636-241500637 on Build GRCh38
rs200004220 Chr.1: 241500572-241500572 on Build GRCh38
rs144131869 Chr.1: 241500602-241500603 on Build GRCh38
rs886046317 Chr.1: 241500603-241500606 on Build GRCh38
rs199911164 Chr.1: 241500604-241500604 on Build GRCh38
rs751518265 Chr.1: 241500596-241500596 on Build GRCh38
rs756369327 Chr.1: 241500605-241500605 on Build GRCh38
rs1022539684 Chr.1: 241500669-241500669 on Build GRCh38
rs757264669 Chr.1: 241500641-241500641 on Build GRCh38
rs1010852437 Chr.1: 241500647-241500647 on Build GRCh38
rs745793347 Chr.1: 241500633-241500634 on Build GRCh38
rs863223989 Chr.1: 241500564-241500564 on Build GRCh38
rs200270634 Chr.1: 241500638-241500638 on Build GRCh38
rs899743834 Chr.1: 241500556-241500556 on Build GRCh38
rs774323879 Chr.1: 241500617-241500618 on Build GRCh38
rs766377516 Chr.1: 241500568-241500568 on Build GRCh38
rs767413280 Chr.1: 241500599-241500599 on Build GRCh38
rs74405673 Chr.1: 241500602-241500602 on Build GRCh38
rs558625619 Chr.1: 241500610-241500610 on Build GRCh38
rs766845458 Chr.1: 241500639-241500640 on Build GRCh38
rs777409218 Chr.1: 241500564-241500564 on Build GRCh38
rs59736583 Chr.1: 241500640-241500641 on Build GRCh38
rs183704256 Chr.1: 241500663-241500663 on Build GRCh38
rs750845779 Chr.1: 241500638-241500639 on Build GRCh38
rs752468689 Chr.1: 241500603-241500603 on Build GRCh38
rs770823245 Chr.1: 241500611-241500612 on Build GRCh38
rs375717640 Chr.1: 241500598-241500598 on Build GRCh38
rs879069854 Chr.1: 241500603-241500604 on Build GRCh38
rs759012809 Chr.1: 241500595-241500604 on Build GRCh38
rs202206776 Chr.1: 241500608-241500608 on Build GRCh38
rs772102092 Chr.1: 241500635-241500636 on Build GRCh38
rs863224009 Chr.1: 241500559-241500559 on Build GRCh38
rs887125405 Chr.1: 241500696-241500696 on Build GRCh38
rs201040163 Chr.1: 241500606-241500606 on Build GRCh38
rs1004260118 Chr.1: 241500737-241500737 on Build GRCh38
rs369072161 Chr.1: 241500601-241500602 on Build GRCh38
rs750898743 Chr.1: 241500594-241500597 on Build GRCh38
rs781528162 Chr.1: 241500603-241500604 on Build GRCh38
rs886046316 Chr.1: 241500594-241500595 on Build GRCh38
rs200926050 Chr.1: 241500595-241500595 on Build GRCh38
rs998472343 Chr.1: 241500657-241500657 on Build GRCh38

View Full Product Details