Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31389034-31389279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB119
Entrez Gene ID
245932
Gene Name
defensin beta 119
Gene Aliases
DEFB-19, DEFB-20, DEFB120, DEFB20, ESC42-RELA, ESC42-RELB
UniGene
Hs.516998
Gene Chromosome Location
Chr.20: 31377163-31390649 on Build GRCh38
Transcripts
NM_001271209.1 NM_153289.3 NM_153323.4 NR_073151.1 NR_073152.1 NR_073153.1 NR_126440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3545017 c.94T>C p.L32L Chr.20: 31389198-31389198 on Build GRCh38
4164663 c.80G>A p.G27E Chr.20: 31389212-31389212 on Build GRCh38
1025543 c.238A>C p.K80Q Chr.20: 31389054-31389054 on Build GRCh38
4399043 c.203G>A p.G68D Chr.20: 31389089-31389089 on Build GRCh38
2888364 c.193G>A p.G65R Chr.20: 31389099-31389099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778891846 Chr.20: 31389203-31389203 on Build GRCh38
rs753147684 Chr.20: 31389099-31389099 on Build GRCh38
rs769136479 Chr.20: 31389235-31389235 on Build GRCh38
rs768593636 Chr.20: 31389134-31389134 on Build GRCh38
rs750663041 Chr.20: 31389128-31389128 on Build GRCh38
rs764291046 Chr.20: 31389109-31389109 on Build GRCh38
rs781630079 Chr.20: 31389120-31389120 on Build GRCh38
rs556128922 Chr.20: 31389083-31389083 on Build GRCh38
rs925275925 Chr.20: 31389175-31389175 on Build GRCh38
rs1052856305 Chr.20: 31389195-31389195 on Build GRCh38
rs772844037 Chr.20: 31389075-31389075 on Build GRCh38
rs1023022403 Chr.20: 31389249-31389249 on Build GRCh38
rs1004592694 Chr.20: 31389140-31389140 on Build GRCh38
rs17327630 Chr.20: 31389146-31389146 on Build GRCh38
rs757381820 Chr.20: 31389118-31389118 on Build GRCh38
rs6120064 Chr.20: 31389208-31389208 on Build GRCh38
rs199988239 Chr.20: 31389204-31389204 on Build GRCh38
rs761285484 Chr.20: 31389074-31389074 on Build GRCh38
rs375754726 Chr.20: 31389155-31389155 on Build GRCh38
rs140963209 Chr.20: 31389145-31389145 on Build GRCh38
rs763475996 Chr.20: 31389107-31389107 on Build GRCh38
rs144611681 Chr.20: 31389243-31389243 on Build GRCh38
rs150443667 Chr.20: 31389150-31389150 on Build GRCh38
rs756307945 Chr.20: 31389180-31389180 on Build GRCh38
rs369081684 Chr.20: 31389202-31389202 on Build GRCh38
rs766114754 Chr.20: 31389184-31389184 on Build GRCh38
rs777217364 Chr.20: 31389206-31389206 on Build GRCh38
rs935309523 Chr.20: 31389179-31389179 on Build GRCh38
rs759872582 Chr.20: 31389080-31389080 on Build GRCh38
rs181386606 Chr.20: 31389161-31389161 on Build GRCh38
rs766664862 Chr.20: 31389185-31389185 on Build GRCh38
rs769942062 Chr.20: 31389198-31389198 on Build GRCh38
rs377193921 Chr.20: 31389242-31389242 on Build GRCh38
rs138332918 Chr.20: 31389168-31389168 on Build GRCh38
rs6120065 Chr.20: 31389209-31389209 on Build GRCh38
rs372464953 Chr.20: 31389149-31389149 on Build GRCh38
rs376955421 Chr.20: 31389113-31389113 on Build GRCh38
rs141277245 Chr.20: 31389131-31389131 on Build GRCh38
rs111324716 Chr.20: 31389162-31389162 on Build GRCh38
rs386813675 Chr.20: 31389208-31389209 on Build GRCh38
rs151140528 Chr.20: 31389067-31389067 on Build GRCh38
rs767724024 Chr.20: 31389244-31389244 on Build GRCh38
rs112304323 Chr.20: 31389132-31389132 on Build GRCh38
rs372699127 Chr.20: 31389215-31389215 on Build GRCh38
rs142077212 Chr.20: 31389213-31389213 on Build GRCh38
rs190164254 Chr.20: 31389156-31389156 on Build GRCh38
rs762049328 Chr.20: 31389167-31389167 on Build GRCh38
rs149638804 Chr.20: 31389197-31389197 on Build GRCh38
rs924495306 Chr.20: 31389159-31389159 on Build GRCh38
rs770078135 Chr.20: 31389151-31389151 on Build GRCh38
rs201738273 Chr.20: 31389177-31389177 on Build GRCh38

View Full Product Details