Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63762426-63762661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
UniGene
Hs.25067
Gene Chromosome Location
Chr.6: 63719980-65707225 on Build GRCh38
Transcripts
NM_001142800.1 NM_001292009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6234845 c.8023_8024delTA p.Y2675fs*14 Chr.6: 63762508-63762509 on Build GRCh38
3160382 c.7934G>A p.C2645Y Chr.6: 63762598-63762598 on Build GRCh38
1080983 c.7930_7931insCAT p.S2643_F2644insS Chr.6: 63762601-63762602 on Build GRCh38
5405700 c.7957G>A p.D2653N Chr.6: 63762575-63762575 on Build GRCh38
1080982 c.7963G>C p.E2655Q Chr.6: 63762569-63762569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748043031 Chr.6: 63762605-63762605 on Build GRCh38
rs996098321 Chr.6: 63762484-63762484 on Build GRCh38
rs749480447 Chr.6: 63762629-63762629 on Build GRCh38
rs919157306 Chr.6: 63762478-63762478 on Build GRCh38
rs374714909 Chr.6: 63762583-63762583 on Build GRCh38
rs757929455 Chr.6: 63762494-63762494 on Build GRCh38
rs950777720 Chr.6: 63762517-63762517 on Build GRCh38
rs527236076 Chr.6: 63762520-63762520 on Build GRCh38
rs918126846 Chr.6: 63762591-63762591 on Build GRCh38
rs976010036 Chr.6: 63762458-63762458 on Build GRCh38
rs909193856 Chr.6: 63762532-63762532 on Build GRCh38
rs527236066 Chr.6: 63762613-63762613 on Build GRCh38
rs779502698 Chr.6: 63762546-63762546 on Build GRCh38
rs769417738 Chr.6: 63762608-63762608 on Build GRCh38
rs778077385 Chr.6: 63762617-63762617 on Build GRCh38
rs751040314 Chr.6: 63762552-63762552 on Build GRCh38
rs780978827 Chr.6: 63762587-63762587 on Build GRCh38
rs898901673 Chr.6: 63762468-63762468 on Build GRCh38
rs879077040 Chr.6: 63762504-63762504 on Build GRCh38
rs1042688038 Chr.6: 63762615-63762615 on Build GRCh38
rs938098897 Chr.6: 63762559-63762559 on Build GRCh38
rs1055183118 Chr.6: 63762571-63762571 on Build GRCh38
rs949510910 Chr.6: 63762595-63762595 on Build GRCh38
rs771244420 Chr.6: 63762636-63762636 on Build GRCh38
rs985211023 Chr.6: 63762529-63762529 on Build GRCh38

View Full Product Details