Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123170723-123170968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM6
Entrez Gene ID
93166
Gene Name
PR domain 6
Gene Aliases
PRISM
UniGene
Hs.135118
Gene Chromosome Location
Chr.5: 123089146-123194266 on Build GRCh38
Transcripts
NM_001136239.1 XM_011543726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336837 c.1264C>T p.P422S Chr.5: 123170876-123170876 on Build GRCh38
5402034 c.1253C>T p.S418F Chr.5: 123170865-123170865 on Build GRCh38
1432709 c.1288T>G p.F430V Chr.5: 123170900-123170900 on Build GRCh38
7385474 c.1231G>A p.A411T Chr.5: 123170843-123170843 on Build GRCh38
163709 c.1333C>G p.Q445E Chr.5: 123170945-123170945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031269045 Chr.5: 123170754-123170754 on Build GRCh38
rs138406807 Chr.5: 123170867-123170867 on Build GRCh38
rs866199814 Chr.5: 123170811-123170811 on Build GRCh38
rs942024099 Chr.5: 123170861-123170861 on Build GRCh38
rs796086351 Chr.5: 123170782-123170782 on Build GRCh38
rs532705349 Chr.5: 123170810-123170810 on Build GRCh38
rs747366998 Chr.5: 123170880-123170880 on Build GRCh38
rs920256314 Chr.5: 123170842-123170842 on Build GRCh38
rs536216409 Chr.5: 123170887-123170887 on Build GRCh38
rs980814235 Chr.5: 123170807-123170807 on Build GRCh38
rs982317028 Chr.5: 123170796-123170796 on Build GRCh38
rs903580424 Chr.5: 123170892-123170892 on Build GRCh38
rs376133768 Chr.5: 123170882-123170882 on Build GRCh38
rs200667727 Chr.5: 123170913-123170913 on Build GRCh38
rs118177028 Chr.5: 123170809-123170809 on Build GRCh38
rs147025859 Chr.5: 123170843-123170843 on Build GRCh38
rs528760231 Chr.5: 123170773-123170773 on Build GRCh38
rs759717526 Chr.5: 123170838-123170838 on Build GRCh38
rs1000548349 Chr.5: 123170898-123170898 on Build GRCh38
rs866297167 Chr.5: 123170923-123170923 on Build GRCh38
rs878951303 Chr.5: 123170801-123170801 on Build GRCh38
rs773395147 Chr.5: 123170761-123170761 on Build GRCh38
rs992345525 Chr.5: 123170762-123170762 on Build GRCh38
rs910507076 Chr.5: 123170862-123170862 on Build GRCh38
rs1036749553 Chr.5: 123170866-123170866 on Build GRCh38
rs1018025633 Chr.5: 123170891-123170891 on Build GRCh38
rs1023772422 Chr.5: 123170781-123170781 on Build GRCh38
rs769825977 Chr.5: 123170907-123170907 on Build GRCh38
rs369520615 Chr.5: 123170886-123170886 on Build GRCh38
rs1006680957 Chr.5: 123170888-123170888 on Build GRCh38
rs776863567 Chr.5: 123170834-123170834 on Build GRCh38
rs547301994 Chr.5: 123170780-123170780 on Build GRCh38
rs201869616 Chr.5: 123170775-123170775 on Build GRCh38

View Full Product Details