Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7089871-7090114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBMXL2
Entrez Gene ID
27288
Gene Name
RNA binding motif protein, X-linked like 2
Gene Aliases
HNRNPG-T, HNRNPGT, HNRPGT
UniGene
Hs.121605
Gene Chromosome Location
Chr.11: 7088934-7091148 on Build GRCh38
Transcripts
NM_014469.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4562008 c.907G>A p.G303R Chr.11: 7090027-7090027 on Build GRCh38
6213416 c.994C>T p.R332W Chr.11: 7090114-7090114 on Build GRCh38
4511801 c.880C>T p.P294S Chr.11: 7090000-7090000 on Build GRCh38
2044439 c.825C>T p.S275S Chr.11: 7089945-7089945 on Build GRCh38
1223403 c.803G>A p.R268H Chr.11: 7089923-7089923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557999059 Chr.11: 7089978-7089978 on Build GRCh38
rs766304493 Chr.11: 7090092-7090092 on Build GRCh38
rs148341004 Chr.11: 7090069-7090069 on Build GRCh38
rs750294343 Chr.11: 7090052-7090052 on Build GRCh38
rs372968144 Chr.11: 7089910-7089910 on Build GRCh38
rs768374330 Chr.11: 7090085-7090085 on Build GRCh38
rs745655284 Chr.11: 7089932-7089932 on Build GRCh38
rs753824653 Chr.11: 7089927-7089927 on Build GRCh38
rs773836383 Chr.11: 7090027-7090029 on Build GRCh38
rs17854944 Chr.11: 7090043-7090043 on Build GRCh38
rs576445396 Chr.11: 7089990-7089990 on Build GRCh38
rs755248402 Chr.11: 7090003-7090003 on Build GRCh38
rs772833571 Chr.11: 7090089-7090089 on Build GRCh38
rs771198132 Chr.11: 7090019-7090019 on Build GRCh38
rs1028042845 Chr.11: 7089946-7089946 on Build GRCh38
rs768689083 Chr.11: 7089949-7089949 on Build GRCh38
rs922197516 Chr.11: 7089995-7089995 on Build GRCh38
rs200655464 Chr.11: 7089958-7089958 on Build GRCh38
rs201798501 Chr.11: 7089894-7089894 on Build GRCh38
rs763685420 Chr.11: 7089988-7089988 on Build GRCh38
rs776242098 Chr.11: 7090044-7090044 on Build GRCh38
rs749844520 Chr.11: 7090017-7090017 on Build GRCh38
rs773591002 Chr.11: 7089959-7089959 on Build GRCh38
rs545441237 Chr.11: 7089997-7089997 on Build GRCh38
rs750183192 Chr.11: 7089998-7089998 on Build GRCh38
rs747271564 Chr.11: 7090026-7090027 on Build GRCh38
rs376176065 Chr.11: 7090006-7090006 on Build GRCh38
rs545267844 Chr.11: 7090054-7090054 on Build GRCh38
rs780160257 Chr.11: 7089944-7089944 on Build GRCh38
rs781418748 Chr.11: 7090066-7090066 on Build GRCh38
rs755121727 Chr.11: 7090063-7090063 on Build GRCh38
rs373146890 Chr.11: 7090077-7090077 on Build GRCh38
rs760472840 Chr.11: 7089979-7089979 on Build GRCh38
rs371314452 Chr.11: 7089904-7089904 on Build GRCh38
rs763252706 Chr.11: 7089974-7089974 on Build GRCh38
rs561845229 Chr.11: 7090083-7090083 on Build GRCh38
rs749747785 Chr.11: 7090072-7090072 on Build GRCh38
rs762738908 Chr.11: 7090090-7090090 on Build GRCh38
rs761743625 Chr.11: 7089993-7089993 on Build GRCh38
rs763740468 Chr.11: 7089921-7089921 on Build GRCh38
rs199683256 Chr.11: 7090029-7090029 on Build GRCh38
rs141524249 Chr.11: 7090088-7090088 on Build GRCh38
rs779355079 Chr.11: 7090078-7090078 on Build GRCh38
rs964630567 Chr.11: 7090005-7090005 on Build GRCh38
rs768429371 Chr.11: 7090041-7090041 on Build GRCh38
rs151210091 Chr.11: 7089905-7089905 on Build GRCh38
rs772446593 Chr.11: 7089901-7089924 on Build GRCh38
rs560600892 Chr.11: 7089891-7089891 on Build GRCh38
rs553443767 Chr.11: 7089903-7089903 on Build GRCh38
rs778123868 Chr.11: 7090016-7090016 on Build GRCh38
rs759166131 Chr.11: 7089908-7089908 on Build GRCh38
rs966227509 Chr.11: 7089963-7089963 on Build GRCh38
rs866482899 Chr.11: 7089937-7089937 on Build GRCh38
rs748177120 Chr.11: 7090007-7090007 on Build GRCh38
rs767111537 Chr.11: 7089909-7089909 on Build GRCh38
rs758104499 Chr.11: 7090000-7090000 on Build GRCh38
rs976228408 Chr.11: 7089968-7089968 on Build GRCh38
rs866874643 Chr.11: 7089982-7089982 on Build GRCh38
rs1010317432 Chr.11: 7089930-7089930 on Build GRCh38
rs751365296 Chr.11: 7090059-7090059 on Build GRCh38
rs757201321 Chr.11: 7089929-7089929 on Build GRCh38
rs201337553 Chr.11: 7090058-7090058 on Build GRCh38
rs777978953 Chr.11: 7090070-7090070 on Build GRCh38
rs143507713 Chr.11: 7089955-7089955 on Build GRCh38
rs141203746 Chr.11: 7089938-7089938 on Build GRCh38
rs770291156 Chr.11: 7090008-7090008 on Build GRCh38
rs138708788 Chr.11: 7089933-7089933 on Build GRCh38
rs924901601 Chr.11: 7090040-7090040 on Build GRCh38
rs766818912 Chr.11: 7089975-7089975 on Build GRCh38
rs780584770 Chr.11: 7089931-7089931 on Build GRCh38
rs779863122 Chr.11: 7090001-7090001 on Build GRCh38
rs777098623 Chr.11: 7089901-7089901 on Build GRCh38
rs933036690 Chr.11: 7090048-7090048 on Build GRCh38
rs753015471 Chr.11: 7090068-7090068 on Build GRCh38
rs774916563 Chr.11: 7090026-7090026 on Build GRCh38
rs761251585 Chr.11: 7090050-7090050 on Build GRCh38
rs192864844 Chr.11: 7089917-7089917 on Build GRCh38
rs549151088 Chr.11: 7090002-7090002 on Build GRCh38
rs199807431 Chr.11: 7089945-7089945 on Build GRCh38

View Full Product Details