Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 71968882-71969143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
UniGene
Hs.485729
Gene Chromosome Location
Chr.6: 71886703-72403143 on Build GRCh38
Transcripts
NM_014989.5 XM_005248688.3 XM_011535604.2 XM_011535611.2 XM_011535615.2 XM_017010516.1 XM_017010517.1 XM_017010518.1 XM_017010519.1 XM_017010520.1 XM_017010521.1 XM_017010522.1 XM_017010523.1 XM_017010524.1 XM_017010525.1 XM_017010526.1 XM_017010527.1 XM_017010528.1 XM_017010529.1 XM_017010530.1 XM_017010531.1 XM_017010532.1 XM_017010533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006550 c.195T>A p.A65A Chr.6: 71969013-71969013 on Build GRCh38
6358909 c.199T>A p.C67S Chr.6: 71969017-71969017 on Build GRCh38
6107111 c.217G>C p.A73P Chr.6: 71969035-71969035 on Build GRCh38
4792883 c.167G>A p.C56Y Chr.6: 71968985-71968985 on Build GRCh38
4480215 c.239C>T p.P80L Chr.6: 71969057-71969057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913674968 Chr.6: 71968973-71968973 on Build GRCh38
rs764932928 Chr.6: 71969050-71969050 on Build GRCh38
rs764174237 Chr.6: 71969041-71969041 on Build GRCh38
rs780463911 Chr.6: 71969056-71969056 on Build GRCh38
rs980106766 Chr.6: 71969029-71969029 on Build GRCh38
rs767344401 Chr.6: 71969015-71969015 on Build GRCh38
rs769887707 Chr.6: 71969090-71969090 on Build GRCh38
rs371681525 Chr.6: 71969099-71969099 on Build GRCh38
rs753365349 Chr.6: 71969045-71969045 on Build GRCh38
rs202189012 Chr.6: 71969100-71969100 on Build GRCh38
rs372890424 Chr.6: 71969052-71969052 on Build GRCh38
rs773456988 Chr.6: 71969004-71969004 on Build GRCh38
rs759206263 Chr.6: 71969008-71969008 on Build GRCh38
rs1028206193 Chr.6: 71968918-71968918 on Build GRCh38
rs376491487 Chr.6: 71969063-71969063 on Build GRCh38
rs200005095 Chr.6: 71968987-71968987 on Build GRCh38
rs561117244 Chr.6: 71968909-71968909 on Build GRCh38
rs781695745 Chr.6: 71969079-71969079 on Build GRCh38
rs746178376 Chr.6: 71968971-71968971 on Build GRCh38
rs569516900 Chr.6: 71968975-71968975 on Build GRCh38
rs755457041 Chr.6: 71969076-71969076 on Build GRCh38
rs528331694 Chr.6: 71968921-71968921 on Build GRCh38
rs926873828 Chr.6: 71969047-71969047 on Build GRCh38
rs765213935 Chr.6: 71969046-71969046 on Build GRCh38
rs748189020 Chr.6: 71969088-71969088 on Build GRCh38
rs778746250 Chr.6: 71968970-71968970 on Build GRCh38
rs760645796 Chr.6: 71969033-71969033 on Build GRCh38
rs532652925 Chr.6: 71968978-71968978 on Build GRCh38
rs772508312 Chr.6: 71968972-71968972 on Build GRCh38
rs756857517 Chr.6: 71969049-71969049 on Build GRCh38
rs755508782 Chr.6: 71968953-71968953 on Build GRCh38
rs775212375 Chr.6: 71969027-71969027 on Build GRCh38
rs752706688 Chr.6: 71968947-71968947 on Build GRCh38
rs766721792 Chr.6: 71968977-71968978 on Build GRCh38
rs769975046 Chr.6: 71969003-71969003 on Build GRCh38
rs756525685 Chr.6: 71968937-71968938 on Build GRCh38
rs759602264 Chr.6: 71968976-71968976 on Build GRCh38
rs185150223 Chr.6: 71969101-71969101 on Build GRCh38
rs756945417 Chr.6: 71968963-71968963 on Build GRCh38
rs180963754 Chr.6: 71968956-71968956 on Build GRCh38
rs995831321 Chr.6: 71968916-71968916 on Build GRCh38
rs367844006 Chr.6: 71969067-71969067 on Build GRCh38
rs758623633 Chr.6: 71969055-71969055 on Build GRCh38
rs374492054 Chr.6: 71968957-71968957 on Build GRCh38

View Full Product Details