Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 29279019-29279258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRU
Entrez Gene ID
10076
Gene Name
protein tyrosine phosphatase, receptor type U
Gene Aliases
FMI, PCP-2, PTP, PTP-J, PTP-PI, PTP-RO, PTPPSI, PTPRO, PTPU2, R-PTP-PSI, R-PTP-U, hPTP-J
UniGene
Hs.19718
Gene Chromosome Location
Chr.1: 29236516-29326813 on Build GRCh38
Transcripts
NM_001195001.1 NM_005704.4 NM_133177.3 NM_133178.3 XM_006710269.2 XM_016999992.1 XM_016999993.1 XM_016999994.1 XM_016999995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7386384 c.1552A>T p.T518S Chr.1: 29279110-29279110 on Build GRCh38
1222703 c.1550T>C p.I517T Chr.1: 29279108-29279108 on Build GRCh38
7386383 c.1552A>T p.T518S Chr.1: 29279110-29279110 on Build GRCh38
4564842 c.1497_1498GG>AA p.E500K Chr.1: 29279055-29279056 on Build GRCh38
1720188 c.1513C>T p.L505F Chr.1: 29279071-29279071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777285251 Chr.1: 29279156-29279156 on Build GRCh38
rs756661606 Chr.1: 29279170-29279170 on Build GRCh38
rs187674909 Chr.1: 29279185-29279185 on Build GRCh38
rs556061908 Chr.1: 29279153-29279153 on Build GRCh38
rs200502248 Chr.1: 29279043-29279043 on Build GRCh38
rs999617593 Chr.1: 29279230-29279230 on Build GRCh38
rs764204163 Chr.1: 29279099-29279099 on Build GRCh38
rs903992683 Chr.1: 29279208-29279208 on Build GRCh38
rs72651203 Chr.1: 29279134-29279134 on Build GRCh38
rs535376460 Chr.1: 29279205-29279205 on Build GRCh38
rs775882102 Chr.1: 29279084-29279084 on Build GRCh38
rs748832366 Chr.1: 29279157-29279157 on Build GRCh38
rs774929231 Chr.1: 29279050-29279050 on Build GRCh38
rs1012442643 Chr.1: 29279175-29279175 on Build GRCh38
rs776833843 Chr.1: 29279101-29279101 on Build GRCh38
rs759990211 Chr.1: 29279052-29279052 on Build GRCh38
rs767261182 Chr.1: 29279138-29279138 on Build GRCh38
rs772512200 Chr.1: 29279053-29279053 on Build GRCh38
rs148293511 Chr.1: 29279088-29279088 on Build GRCh38
rs895376958 Chr.1: 29279166-29279166 on Build GRCh38
rs572392319 Chr.1: 29279229-29279229 on Build GRCh38
rs751442773 Chr.1: 29279126-29279126 on Build GRCh38
rs907538668 Chr.1: 29279090-29279090 on Build GRCh38
rs761797398 Chr.1: 29279104-29279104 on Build GRCh38
rs150842094 Chr.1: 29279111-29279111 on Build GRCh38
rs372895985 Chr.1: 29279150-29279150 on Build GRCh38

View Full Product Details