Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7927936-7928181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM44
Entrez Gene ID
10469
Gene Name
translocase of inner mitochondrial membrane 44
Gene Aliases
TIM44
UniGene
Hs.465784
Gene Chromosome Location
Chr.19: 7926718-7943823 on Build GRCh38
Transcripts
NM_006351.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5454792 c.1039-7A>G p.? Chr.19: 7928173-7928173 on Build GRCh38
1305316 c.1117G>A p.D373N Chr.19: 7928088-7928088 on Build GRCh38
5857129 c.1066C>T p.Q356* Chr.19: 7928139-7928139 on Build GRCh38
4821366 c.1125C>T p.V375V Chr.19: 7928080-7928080 on Build GRCh38
7145480 c.1065C>G p.I355M Chr.19: 7928140-7928140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369918101 Chr.19: 7928080-7928080 on Build GRCh38
rs373424642 Chr.19: 7928066-7928066 on Build GRCh38
rs766510463 Chr.19: 7928084-7928084 on Build GRCh38
rs34425383 Chr.19: 7928074-7928074 on Build GRCh38
rs944083338 Chr.19: 7928041-7928041 on Build GRCh38
rs925116382 Chr.19: 7927995-7927995 on Build GRCh38
rs756120261 Chr.19: 7928144-7928144 on Build GRCh38
rs912509228 Chr.19: 7928149-7928149 on Build GRCh38
rs1013292791 Chr.19: 7928082-7928082 on Build GRCh38
rs767657869 Chr.19: 7928090-7928090 on Build GRCh38
rs767606481 Chr.19: 7928012-7928012 on Build GRCh38
rs779912806 Chr.19: 7928140-7928140 on Build GRCh38
rs201680343 Chr.19: 7928053-7928053 on Build GRCh38
rs771796001 Chr.19: 7928065-7928065 on Build GRCh38
rs149334518 Chr.19: 7928132-7928132 on Build GRCh38
rs912456987 Chr.19: 7928017-7928017 on Build GRCh38
rs892056128 Chr.19: 7928034-7928034 on Build GRCh38
rs200747180 Chr.19: 7928088-7928088 on Build GRCh38
rs149261738 Chr.19: 7928050-7928050 on Build GRCh38
rs779947854 Chr.19: 7928037-7928037 on Build GRCh38
rs150088116 Chr.19: 7928102-7928102 on Build GRCh38
rs764339222 Chr.19: 7928101-7928101 on Build GRCh38
rs774364066 Chr.19: 7928086-7928086 on Build GRCh38
rs969542639 Chr.19: 7927965-7927965 on Build GRCh38
rs756615799 Chr.19: 7928032-7928032 on Build GRCh38
rs138623078 Chr.19: 7928125-7928125 on Build GRCh38
rs761625456 Chr.19: 7928077-7928077 on Build GRCh38
rs898578423 Chr.19: 7927962-7927962 on Build GRCh38
rs200515886 Chr.19: 7928078-7928078 on Build GRCh38
rs11542187 Chr.19: 7928091-7928091 on Build GRCh38
rs759766479 Chr.19: 7928089-7928089 on Build GRCh38
rs745541892 Chr.19: 7928056-7928056 on Build GRCh38
rs529915396 Chr.19: 7927960-7927960 on Build GRCh38
rs539846855 Chr.19: 7927992-7927992 on Build GRCh38
rs745315611 Chr.19: 7928103-7928103 on Build GRCh38
rs374813553 Chr.19: 7928042-7928042 on Build GRCh38
rs199532231 Chr.19: 7928030-7928030 on Build GRCh38
rs768825417 Chr.19: 7928160-7928160 on Build GRCh38
rs756525735 Chr.19: 7928098-7928098 on Build GRCh38
rs768324341 Chr.19: 7928068-7928068 on Build GRCh38
rs746638693 Chr.19: 7928157-7928157 on Build GRCh38
rs770519947 Chr.19: 7928052-7928052 on Build GRCh38
rs566310395 Chr.19: 7928028-7928028 on Build GRCh38
rs987960603 Chr.19: 7928113-7928113 on Build GRCh38
rs935113154 Chr.19: 7928000-7928000 on Build GRCh38
rs1053269923 Chr.19: 7928018-7928018 on Build GRCh38

View Full Product Details