Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46310102-46310373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UQCRH
Entrez Gene ID
7388
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases
QCR6, UQCR8
UniGene
Hs.481571
Gene Chromosome Location
Chr.1: 46303613-46316777 on Build GRCh38
Transcripts
NM_001297565.1 NM_001297566.1 NM_006004.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2191183 c.230C>T p.A77V Chr.1: 46310303-46310303 on Build GRCh38
2191182 c.178C>T p.R60* Chr.1: 46310251-46310251 on Build GRCh38
6817721 c.111C>T p.C37C Chr.1: 46310184-46310184 on Build GRCh38
6284361 c.149T>A p.L50Q Chr.1: 46310222-46310222 on Build GRCh38
2191181 c.166C>T p.R56C Chr.1: 46310239-46310239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933121886 Chr.1: 46310253-46310253 on Build GRCh38
rs773786491 Chr.1: 46310125-46310125 on Build GRCh38
rs147731563 Chr.1: 46310185-46310185 on Build GRCh38
rs34813470 Chr.1: 46310224-46310224 on Build GRCh38
rs761257439 Chr.1: 46310138-46310138 on Build GRCh38
rs928398501 Chr.1: 46310163-46310165 on Build GRCh38
rs752881436 Chr.1: 46310261-46310261 on Build GRCh38
rs747396129 Chr.1: 46310215-46310215 on Build GRCh38
rs146621405 Chr.1: 46310213-46310213 on Build GRCh38
rs199979488 Chr.1: 46310135-46310135 on Build GRCh38
rs778268018 Chr.1: 46310212-46310212 on Build GRCh38
rs769377443 Chr.1: 46310316-46310316 on Build GRCh38
rs745691114 Chr.1: 46310227-46310228 on Build GRCh38
rs761559224 Chr.1: 46310169-46310169 on Build GRCh38
rs778680583 Chr.1: 46310197-46310204 on Build GRCh38
rs919028614 Chr.1: 46310252-46310252 on Build GRCh38
rs11556324 Chr.1: 46310210-46310210 on Build GRCh38
rs189114560 Chr.1: 46310233-46310233 on Build GRCh38
rs757022954 Chr.1: 46310172-46310173 on Build GRCh38
rs531150957 Chr.1: 46310298-46310298 on Build GRCh38
rs148934256 Chr.1: 46310262-46310262 on Build GRCh38
rs748384022 Chr.1: 46310205-46310205 on Build GRCh38
rs1053711515 Chr.1: 46310281-46310281 on Build GRCh38
rs139334642 Chr.1: 46310170-46310170 on Build GRCh38
rs767749634 Chr.1: 46310240-46310240 on Build GRCh38
rs766320025 Chr.1: 46310247-46310247 on Build GRCh38
rs746368189 Chr.1: 46310288-46310288 on Build GRCh38
rs549566186 Chr.1: 46310304-46310304 on Build GRCh38
rs751232369 Chr.1: 46310159-46310159 on Build GRCh38
rs753379099 Chr.1: 46310131-46310132 on Build GRCh38
rs370156737 Chr.1: 46310268-46310268 on Build GRCh38
rs554426072 Chr.1: 46310271-46310271 on Build GRCh38
rs755064731 Chr.1: 46310193-46310193 on Build GRCh38
rs148062188 Chr.1: 46310277-46310277 on Build GRCh38
rs753743917 Chr.1: 46310248-46310248 on Build GRCh38
rs780579191 Chr.1: 46310350-46310350 on Build GRCh38
rs368398364 Chr.1: 46310327-46310327 on Build GRCh38
rs376054810 Chr.1: 46310292-46310292 on Build GRCh38
rs761472665 Chr.1: 46310227-46310227 on Build GRCh38
rs751759087 Chr.1: 46310325-46310325 on Build GRCh38
rs762474674 Chr.1: 46310219-46310219 on Build GRCh38
rs746293977 Chr.1: 46310338-46310338 on Build GRCh38
rs779146646 Chr.1: 46310257-46310257 on Build GRCh38
rs772506078 Chr.1: 46310208-46310208 on Build GRCh38
rs771218841 Chr.1: 46310276-46310276 on Build GRCh38
rs201735339 Chr.1: 46310139-46310139 on Build GRCh38
rs775328081 Chr.1: 46310331-46310331 on Build GRCh38
rs773841151 Chr.1: 46310296-46310296 on Build GRCh38
rs777050301 Chr.1: 46310218-46310218 on Build GRCh38
rs755039048 Chr.1: 46310254-46310254 on Build GRCh38
rs775056917 Chr.1: 46310145-46310145 on Build GRCh38
rs772789538 Chr.1: 46310300-46310300 on Build GRCh38
rs756613690 Chr.1: 46310345-46310345 on Build GRCh38
rs377206046 Chr.1: 46310143-46310143 on Build GRCh38
rs144503839 Chr.1: 46310148-46310148 on Build GRCh38
rs201496001 Chr.1: 46310317-46310317 on Build GRCh38
rs11556325 Chr.1: 46310178-46310178 on Build GRCh38
rs112106837 Chr.1: 46310319-46310319 on Build GRCh38
rs750228344 Chr.1: 46310239-46310239 on Build GRCh38
rs779234901 Chr.1: 46310201-46310201 on Build GRCh38
rs756068720 Chr.1: 46310184-46310184 on Build GRCh38
rs765331614 Chr.1: 46310310-46310310 on Build GRCh38
rs763711552 Chr.1: 46310150-46310150 on Build GRCh38
rs181971906 Chr.1: 46310303-46310303 on Build GRCh38
rs370395351 Chr.1: 46310217-46310217 on Build GRCh38

View Full Product Details