Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 44934948-44935177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM72
Entrez Gene ID
643236
Gene Name
transmembrane protein 72
Gene Aliases
C10orf127, KSP37
UniGene
Hs.292788
Gene Chromosome Location
Chr.10: 44911307-44937010 on Build GRCh38
Transcripts
NM_001123376.2 XM_011540069.2 XM_011540070.2 XM_011540071.2 NR_033842.1

Gene Symbol
TMEM72-AS1
Entrez Gene ID
220980
Gene Name
TMEM72 antisense RNA 1
Gene Aliases
-
UniGene
Hs.568801
Gene Chromosome Location
Chr.10: 44811024-44959689 on Build GRCh38
Transcripts
NM_001123376.2 XM_011540069.2 XM_011540070.2 XM_011540071.2 NR_033842.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4847083 c.733G>C p.E245Q Chr.10: 44935039-44935039 on Build GRCh38
5370643 c.790C>T p.L264F Chr.10: 44935096-44935096 on Build GRCh38
4734602 c.697C>T p.P233S Chr.10: 44935003-44935003 on Build GRCh38
6482674 c.745A>G p.T249A Chr.10: 44935051-44935051 on Build GRCh38
4557724 c.739G>A p.E247K Chr.10: 44935045-44935045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007979919 Chr.10: 44935108-44935108 on Build GRCh38
rs762751980 Chr.10: 44935027-44935027 on Build GRCh38
rs755886971 Chr.10: 44934986-44934986 on Build GRCh38
rs758456832 Chr.10: 44935069-44935069 on Build GRCh38
rs1050584980 Chr.10: 44935091-44935091 on Build GRCh38
rs777410637 Chr.10: 44934991-44934991 on Build GRCh38
rs201585558 Chr.10: 44935045-44935045 on Build GRCh38
rs760728564 Chr.10: 44935122-44935122 on Build GRCh38
rs764493556 Chr.10: 44935082-44935082 on Build GRCh38
rs201705501 Chr.10: 44935158-44935158 on Build GRCh38
rs968635967 Chr.10: 44935153-44935153 on Build GRCh38
rs777128580 Chr.10: 44935026-44935026 on Build GRCh38
rs771656318 Chr.10: 44935111-44935111 on Build GRCh38
rs774094759 Chr.10: 44935102-44935102 on Build GRCh38
rs200252955 Chr.10: 44935128-44935128 on Build GRCh38
rs371286835 Chr.10: 44935065-44935065 on Build GRCh38
rs940858274 Chr.10: 44935013-44935013 on Build GRCh38
rs374225789 Chr.10: 44935094-44935094 on Build GRCh38
rs767417890 Chr.10: 44934975-44934975 on Build GRCh38
rs576944254 Chr.10: 44935136-44935136 on Build GRCh38
rs780237081 Chr.10: 44935138-44935138 on Build GRCh38
rs920366417 Chr.10: 44935050-44935050 on Build GRCh38
rs908098295 Chr.10: 44935011-44935011 on Build GRCh38
rs781748745 Chr.10: 44935155-44935155 on Build GRCh38
rs377099938 Chr.10: 44934979-44934979 on Build GRCh38
rs371052431 Chr.10: 44935036-44935036 on Build GRCh38
rs759402619 Chr.10: 44935084-44935084 on Build GRCh38
rs372665382 Chr.10: 44935012-44935012 on Build GRCh38
rs754975609 Chr.10: 44934972-44934972 on Build GRCh38
rs759073093 Chr.10: 44935042-44935042 on Build GRCh38
rs750694378 Chr.10: 44935137-44935137 on Build GRCh38
rs777146968 Chr.10: 44935095-44935095 on Build GRCh38
rs12249378 Chr.10: 44935157-44935157 on Build GRCh38
rs764020711 Chr.10: 44935055-44935055 on Build GRCh38
rs770707129 Chr.10: 44934973-44934973 on Build GRCh38
rs190226613 Chr.10: 44935141-44935141 on Build GRCh38
rs17417442 Chr.10: 44935014-44935014 on Build GRCh38
rs759085092 Chr.10: 44935105-44935105 on Build GRCh38
rs751100915 Chr.10: 44934967-44934967 on Build GRCh38
rs776467255 Chr.10: 44935011-44935011 on Build GRCh38
rs372932789 Chr.10: 44934995-44934995 on Build GRCh38
rs376784361 Chr.10: 44935058-44935058 on Build GRCh38
rs747357552 Chr.10: 44935015-44935015 on Build GRCh38
rs769102816 Chr.10: 44935019-44935019 on Build GRCh38
rs774860990 Chr.10: 44935121-44935121 on Build GRCh38
rs767375435 Chr.10: 44935044-44935044 on Build GRCh38
rs755976958 Chr.10: 44935052-44935052 on Build GRCh38
rs755512615 Chr.10: 44935150-44935150 on Build GRCh38
rs369250579 Chr.10: 44934998-44934998 on Build GRCh38
rs201970986 Chr.10: 44935123-44935123 on Build GRCh38
rs766071234 Chr.10: 44935029-44935029 on Build GRCh38
rs779964969 Chr.10: 44935072-44935072 on Build GRCh38
rs369667578 Chr.10: 44934994-44934994 on Build GRCh38
rs753619992 Chr.10: 44935063-44935063 on Build GRCh38
rs368153016 Chr.10: 44935101-44935101 on Build GRCh38
rs571432690 Chr.10: 44935087-44935087 on Build GRCh38
rs940728598 Chr.10: 44935048-44935048 on Build GRCh38
rs141426203 Chr.10: 44935064-44935064 on Build GRCh38
rs747616132 Chr.10: 44935078-44935078 on Build GRCh38

View Full Product Details