Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 100448327-100448552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIM1
Entrez Gene ID
6492
Gene Name
single-minded family bHLH transcription factor 1
Gene Aliases
bHLHe14
UniGene
Hs.520293
Gene Chromosome Location
Chr.6: 100385009-100464929 on Build GRCh38
Transcripts
NM_005068.2 XM_005267100.2 XM_011536072.2 XM_017011197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6044006 c.706G>A p.A236T Chr.6: 100448516-100448516 on Build GRCh38
1071585 c.705C>T p.R235R Chr.6: 100448517-100448517 on Build GRCh38
318026 c.675C>A p.I225I Chr.6: 100448547-100448547 on Build GRCh38
1487070 c.715G>C p.D239H Chr.6: 100448507-100448507 on Build GRCh38
3142515 c.684C>T p.H228H Chr.6: 100448538-100448538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551202042 Chr.6: 100448400-100448400 on Build GRCh38
rs945718396 Chr.6: 100448364-100448364 on Build GRCh38
rs780269717 Chr.6: 100448498-100448498 on Build GRCh38
rs1045382253 Chr.6: 100448365-100448365 on Build GRCh38
rs756054020 Chr.6: 100448472-100448472 on Build GRCh38
rs375331827 Chr.6: 100448380-100448381 on Build GRCh38
rs973397881 Chr.6: 100448407-100448407 on Build GRCh38
rs762400046 Chr.6: 100448442-100448442 on Build GRCh38
rs765989967 Chr.6: 100448444-100448444 on Build GRCh38
rs765705820 Chr.6: 100448429-100448429 on Build GRCh38
rs752577075 Chr.6: 100448465-100448465 on Build GRCh38
rs376533606 Chr.6: 100448509-100448509 on Build GRCh38
rs201781365 Chr.6: 100448510-100448510 on Build GRCh38
rs3830139 Chr.6: 100448413-100448413 on Build GRCh38
rs750478646 Chr.6: 100448487-100448487 on Build GRCh38
rs997223762 Chr.6: 100448347-100448347 on Build GRCh38
rs747219514 Chr.6: 100448506-100448506 on Build GRCh38
rs767411359 Chr.6: 100448464-100448464 on Build GRCh38
rs759317253 Chr.6: 100448455-100448455 on Build GRCh38
rs971796732 Chr.6: 100448425-100448425 on Build GRCh38
rs886060897 Chr.6: 100448469-100448469 on Build GRCh38
rs758968335 Chr.6: 100448385-100448385 on Build GRCh38
rs950967845 Chr.6: 100448433-100448433 on Build GRCh38
rs1029155312 Chr.6: 100448362-100448362 on Build GRCh38
rs377605969 Chr.6: 100448447-100448447 on Build GRCh38
rs758417431 Chr.6: 100448493-100448493 on Build GRCh38
rs781506752 Chr.6: 100448513-100448513 on Build GRCh38
rs1055460867 Chr.6: 100448380-100448380 on Build GRCh38
rs537591799 Chr.6: 100448371-100448371 on Build GRCh38
rs745406086 Chr.6: 100448431-100448431 on Build GRCh38
rs962931303 Chr.6: 100448399-100448399 on Build GRCh38
rs772728888 Chr.6: 100448441-100448441 on Build GRCh38
rs777597365 Chr.6: 100448476-100448476 on Build GRCh38
rs764985586 Chr.6: 100448394-100448394 on Build GRCh38
rs3213541 Chr.6: 100448367-100448367 on Build GRCh38
rs769255246 Chr.6: 100448458-100448458 on Build GRCh38

View Full Product Details