Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16863597-16863870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIN3B
Entrez Gene ID
23309
Gene Name
SIN3 transcription regulator family member B
Gene Aliases
-
UniGene
Hs.13999
Gene Chromosome Location
Chr.19: 16829387-16880355 on Build GRCh38
Transcripts
NM_001297595.1 NM_001297597.1 NM_015260.3 XM_006722704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6279554 c.1437G>A p.P479P Chr.19: 16863754-16863754 on Build GRCh38
7136734 c.1412C>T p.T471M Chr.19: 16863729-16863729 on Build GRCh38
5028518 c.1404G>T p.E468D Chr.19: 16863721-16863721 on Build GRCh38
4580760 c.1473C>A p.R491R Chr.19: 16863790-16863790 on Build GRCh38
5139704 c.1479+1_1479+2delGT p.? Chr.19: 16863797-16863798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776639335 Chr.19: 16863706-16863706 on Build GRCh38
rs200011485 Chr.19: 16863817-16863817 on Build GRCh38
rs780267365 Chr.19: 16863668-16863668 on Build GRCh38
rs763313414 Chr.19: 16863748-16863748 on Build GRCh38
rs747901929 Chr.19: 16863673-16863673 on Build GRCh38
rs746545500 Chr.19: 16863688-16863688 on Build GRCh38
rs908387234 Chr.19: 16863660-16863660 on Build GRCh38
rs746657713 Chr.19: 16863753-16863753 on Build GRCh38
rs12461731 Chr.19: 16863730-16863730 on Build GRCh38
rs754131940 Chr.19: 16863816-16863816 on Build GRCh38
rs375392788 Chr.19: 16863651-16863651 on Build GRCh38
rs907257212 Chr.19: 16863785-16863785 on Build GRCh38
rs140663662 Chr.19: 16863793-16863793 on Build GRCh38
rs567346444 Chr.19: 16863822-16863822 on Build GRCh38
rs190417952 Chr.19: 16863625-16863625 on Build GRCh38
rs751788314 Chr.19: 16863758-16863758 on Build GRCh38
rs771683996 Chr.19: 16863674-16863674 on Build GRCh38
rs187490262 Chr.19: 16863623-16863623 on Build GRCh38
rs535323114 Chr.19: 16863810-16863810 on Build GRCh38
rs775114267 Chr.19: 16863734-16863734 on Build GRCh38
rs922335180 Chr.19: 16863643-16863643 on Build GRCh38
rs371358286 Chr.19: 16863646-16863646 on Build GRCh38
rs367560628 Chr.19: 16863644-16863644 on Build GRCh38
rs769592244 Chr.19: 16863837-16863837 on Build GRCh38
rs770392179 Chr.19: 16863691-16863691 on Build GRCh38
rs777313390 Chr.19: 16863675-16863675 on Build GRCh38
rs200476140 Chr.19: 16863650-16863650 on Build GRCh38
rs370397656 Chr.19: 16863757-16863757 on Build GRCh38
rs894438012 Chr.19: 16863744-16863744 on Build GRCh38
rs964986081 Chr.19: 16863850-16863850 on Build GRCh38
rs746600691 Chr.19: 16863821-16863821 on Build GRCh38
rs756512601 Chr.19: 16863667-16863667 on Build GRCh38
rs150931359 Chr.19: 16863754-16863754 on Build GRCh38
rs758683680 Chr.19: 16863645-16863645 on Build GRCh38
rs376687511 Chr.19: 16863823-16863823 on Build GRCh38
rs756574150 Chr.19: 16863799-16863799 on Build GRCh38
rs767438597 Chr.19: 16863790-16863790 on Build GRCh38
rs201985254 Chr.19: 16863826-16863826 on Build GRCh38
rs775445448 Chr.19: 16863840-16863840 on Build GRCh38
rs369985112 Chr.19: 16863733-16863733 on Build GRCh38
rs761987982 Chr.19: 16863784-16863784 on Build GRCh38
rs764878194 Chr.19: 16863641-16863641 on Build GRCh38
rs374859820 Chr.19: 16863636-16863636 on Build GRCh38
rs750828265 Chr.19: 16863659-16863659 on Build GRCh38
rs780509139 Chr.19: 16863804-16863804 on Build GRCh38
rs183061197 Chr.19: 16863825-16863825 on Build GRCh38
rs746855389 Chr.19: 16863648-16863648 on Build GRCh38

View Full Product Details